Skip to Content
Merck
All Photos(3)

Key Documents

B67201

Sigma-Aldrich

1-Bromo-4-fluorobenzene

99%

Synonym(s):

1-Fluoro-4-bromobenzene, 4-Bromo-1-fluorobenzene, 4-Fluoro-1-bromobenzene, p-Fluorophenyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$32.90
100 G
$80.80
500 G
$232.00

$32.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$32.90
100 G
$80.80
500 G
$232.00

About This Item

Linear Formula:
BrC6H4F
CAS Number:
Molecular Weight:
175.00
Beilstein:
774102
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$32.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.527 (lit.)

bp

150 °C (lit.)

mp

−16 °C (lit.)

density

1.593 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Br)cc1

InChI

1S/C6H4BrF/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

AITNMTXHTIIIBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-4-fluorobenzene is commonly used in the Suzuki-Miyaura coupling reaction to form carbon-carbon bonds.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki-Miyaura CC coupling reactions catalyzed by supported Pd nanoparticles for the preparation of fluorinated biphenyl derivatives
Erami, R et al.
Catalysts, 7(3), 76-76 (2017)
Qian Luo et al.
Journal of chromatography. A, 1365, 45-53 (2014-09-27)
When iodide and natural organic matter are present in raw water, the formation of iodo-trihalomethanes (Iodo-THMs), haloacetonitriles (HANs), and halonitromethanes (HNMs) pose a potential health risk because they have been reported to be more toxic than their brominated or chlorinated
Michael Phillips et al.
Health physics, 108(5), 538-546 (2015-03-27)
There is widespread interest in the development of tools to estimate radiation exposures. Exhaled breath provides a novel matrix for assessing biomarkers that could be correlated with exposures. The use of exhaled breath for estimating radiation exposure is warranted, as
Jae Kwak et al.
Journal of separation science, 38(14), 2463-2469 (2015-05-07)
Volatile organic compounds were collected and analyzed from a variety of indoor and outdoor air samples to test whether human-derived compounds can be readily detected in the air and if they can be associated with human occupancy or presence. Compounds
Christoph Hutzler et al.
Archives of toxicology, 88(7), 1295-1308 (2014-06-25)
Electronic (e-)cigarettes have emerged in recent years as putative alternative to conventional tobacco cigarettes. These products do not contain typical carcinogens that are present in tobacco smoke, due to the lack of combustion. However, besides nicotine, hazards can also arise

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service