Skip to Content
Merck
All Photos(1)

Key Documents

279463

Sigma-Aldrich

Dimethyl-d6-amine hydrochloride

99 atom % D

Synonym(s):

Deuterated dimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$307.00
5 G
$1,130.00

$307.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$307.00
5 G
$1,130.00

About This Item

Linear Formula:
(CD3)2NH · HCl
CAS Number:
Molecular Weight:
87.58
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$307.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Assay

≥99% (ion chromatography)

form

solid

mp

170-173 °C (lit.)

mass shift

M+6

storage temp.

room temp

SMILES string

Cl.[2H]C([2H])([2H])NC([2H])([2H])[2H]

InChI

1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H/i1D3,2D3;

InChI key

IQDGSYLLQPDQDV-TXHXQZCNSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Rok et al.
Dalton transactions (Cambridge, England : 2003), 48(13), 4190-4202 (2019-03-02)
We have synthesised and characterised a novel organic-inorganic hybrid crystal, [(CH3)2NH2]2KCr(CN)6. The thermal DSC, TMA, DTG and DTA analyses indicate two solid-to-solid structural phase transitions (PTs). According to the X-ray diffraction experiments, the first PT at 220 K is isostructural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service