Skip to Content
Merck
All Photos(3)

Documents

149357

Sigma-Aldrich

2,5-Dihydroxybenzoic acid

98%

Synonym(s):

2,5-DHBA, DHB, Gentisic acid, Hydroquinonecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
2209119
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

204-208 °C (lit.)

SMILES string

OC(=O)c1cc(O)ccc1O

InChI

1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

WXTMDXOMEHJXQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Aspirin metabolite.

Application

2,5-Dihydroxybenzoic acid (2,5-DHBA) can be used:
  • As a matrix in the MALDI-TOF mass spectrometry analysis of the poly(styrene-co-pentafluorostyrene) copolymers.
  • To prepare pharmacologically important 2,5-DBHA-gelatin conjugate, which can act as a polymer for the development of drug formulations.
  • As a tridentate ligand to synthesize complexes of molybdenum by reacting with (PPh4)4Mo8O26.
  • As a starting material in the total synthesis of landomycin A, an angucycline antibiotic.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evgenia Tisdale et al.
Analytica chimica acta, 808, 151-162 (2013-12-29)
The influence of the sample preparation parameters (the choice of the matrix, matrix:analyte ratio, salt:analyte ratio) was investigated and optimal conditions were established for the MALDI time-of-flight mass spectrometry analysis of the poly(styrene-co-pentafluorostyrene) copolymers. These were synthesized by atom transfer
Matrix-assisted laser desorption/ionization sample preparation optimization for structural characterization of poly (styrene-co-pentafluorostyrene) copolymers
Tisdale E, et al.
Analytica Chimica Acta, 808, 151-162 (2014)
Polynuclear oxomolybdenum (VI) complexes of dihydroxybenzoic acids: Synthesis, spectroscopic and structure characterization of a tetranuclear catecholato-type coordinated 2, 3-dihydroxybenzoate and a novel tridentate salicylato-type coordinated 2, 5-dihydroxybenzoate trinuclear complex
Litos C, et al.
Polyhedron, 25(6), 1337-1347 (2006)
A 2, 5-Dihydroxybenzoic acid-gelatin conjugate: the synthesis, antiviral activity and mechanism of antiviral action against two alphaherpesviruses
Lisov A, et al.
Viruses, 7(10), 5343-5360 (2015)
Matthew T Briggs et al.
Rapid communications in mass spectrometry : RCM, 31(10), 825-841 (2017-03-09)
Matrix-assisted laser desorption/ionization mass spectrometry imaging (MALDI-MSI) of the proteome of a tissue has been an established technique for the past decade. In the last few years, MALDI-MSI of the N-glycome has emerged as a novel MALDI-MSI technique. To assess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service