Skip to Content
Merck
All Photos(1)

Key Documents

78069

Supelco

2,5-Dihydroxybenzoic acid

analytical standard

Synonym(s):

2,5-DHBA, DHB, Gentisic acid, Hydroquinonecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$108.00

$108.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
$108.00

About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
2209119
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

$108.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

204-208 °C (lit.)

application(s)

pharmaceutical

format

neat

SMILES string

OC(=O)c1cc(O)ccc1O

InChI

1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

WXTMDXOMEHJXQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Dihydroxybenzoic acid may be used as a reference standard in the determination of 2,5-dihydroxybenzoic acid in biopsy samples using high-performance liquid chromatography coupled with electrochemical detector.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurent Salphati et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(7), 1110-1116 (2014-04-24)
Glioblastoma multiforme (GBM) is the most common primary brain tumor in adults, and the limited available treatment options have not meaningfully impacted patient survival in the past decades. Such poor outcomes can be at least partly attributed to the inability
G Palumbo et al.
Journal of pharmaceutical and biomedical analysis, 14(1-2), 175-180 (1995-12-01)
A high-performance liquid chromatographic method for the simultaneous determination of 5-aminosalicylic acid (5-ASA), acetyl-5-aminosalicylic acid (Ac-5-ASA) and 2,5-dihydroxybenzoic acid (5-HSA) in human endoscopic intestinal biopsy with electrochemical detection has been developed and validated. A liquid-liquid extraction procedure was used to
H Ma et al.
Oncogene, 34(6), 726-740 (2014-02-18)
Aberrant cell surface sialylation patterns have been shown to correlate with tumor progression and metastasis. However, the role of sialylation regulation of cancer multidrug resistance (MDR) remains poorly understood. This study investigated sialylation in modification on MDR in acute myeloid
Shadi Toghi Eshghi et al.
ACS chemical biology, 9(9), 2149-2156 (2014-07-17)
Aberrant glycosylation is associated with most of the diseases. Direct imaging and profiling of N-glycans on tissue sections can reveal tissue-specific and/or disease-associated N-glycans, which not only could serve as molecular signatures for diagnosis but also shed light on the
S A Ponomarenko et al.
Faraday discussions, 174, 313-339 (2014-10-04)
This contribution describes recent progress in the design, synthesis and properties of solution-processible star-shaped oligomers and their application in organic photovoltaics. Even though alternative chemistry has been used to design such oligomers, the most successful approach is based on a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service