Skip to Content
Merck
All Photos(1)

Documents

8.41745

Sigma-Aldrich

2,5-Dihydroxybenzoic acid

for synthesis

Synonym(s):

2,5-Dihydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
207-718-5
NACRES:
NA.22

Quality Level

Assay

≥99.0% (HPLC)

form

powder

mp

202-206 °C

storage temp.

2-30°C

InChI

1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

WXTMDXOMEHJXQO-UHFFFAOYSA-N

Related Categories

Application

2,5-Dihydroxybenzoic acid for synthesis. CAS 490-79-9, molar mass 154.12 g/mol.

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 200 °C
Melting range (upper value): ≤ 206 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service