Skip to Content
Merck
All Photos(1)

Key Documents

541060

Sigma-Aldrich

Ethyl 2,5-dihydroxybenzoate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$72.70

$72.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$72.70

About This Item

Linear Formula:
(HO)2C6H3CO2CH2CH3
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$72.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

77-81 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cc(O)ccc1O

InChI

1S/C9H10O4/c1-2-13-9(12)7-5-6(10)3-4-8(7)11/h3-5,10-11H,2H2,1H3

InChI key

GCUPAENRSCPHBM-UHFFFAOYSA-N

General description

Ethyl 2,5-dihydroxybenzoate, also known as ethyl gentisate, is an aromatic ester. It can be synthesized from 2,5-dihydroxybenzoic acid by esterification process with absolute ethanol.[1]

Application

Ethyl 2,5-dihydroxybenzoate may be used for developing fourth generation acid dendrons.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembled lanthanide-cored dendrimer complexes: enhancement of the luminescence properties of lanthanide ions through site-isolation and antenna effects.
Kawa M and Frechet JMJ.
Chemistry of Materials, 10(1), 286-296 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service