Skip to Content
Merck
All Photos(1)

Key Documents

125113

Sigma-Aldrich

2-Chloro-5-nitrobenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$74.30

$74.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$74.30

About This Item

Linear Formula:
ClC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
201.56
Beilstein:
1877474
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

165-168 °C (lit.)

functional group

carboxylic acid
chloro
nitro

SMILES string

OC(=O)c1cc(ccc1Cl)[N+]([O-])=O

InChI

1S/C7H4ClNO4/c8-6-2-1-4(9(12)13)3-5(6)7(10)11/h1-3H,(H,10,11)

InChI key

QUEKGYQTRJVEQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-5-nitrobenzoic acid undergoes microwave-assisted, regioselective amination reaction with aliphatic and aromatic amines to yield N-substituted 5-nitroanthranilic acid derivatives[1]. It acts as ligand and forms a red luminescent one dimensional coordination polymer with Eu(III)[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

2-Chloro-5-nitrobenzoato complexes of Eu (III) and Tb (III)-A 1D coordination polymer and enhanced solution luminescence.
Viswanathan S and Bettencourt-Dias A.
Inorganic Chemistry Communications, 9(5), 444-448 (2006)
Younis Baqi et al.
The Journal of organic chemistry, 72(15), 5908-5911 (2007-06-26)
The synthesis of N-substituted 5-nitroanthranilic acid derivatives 3a-w was achieved by a new, mild, microwave-assisted, regioselective amination reaction of 5-nitro-2-chlorobenzoic acid (1a) with a diverse range of aliphatic and aromatic amines 2a-w without added solvent or catalyst. Up to >99%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service