Key Documents
57331
Supelco
SPME Fiber Holder
greener alternative
for use with Non-CTC CombiPAL Type Autosamplers
About This Item
UNSPSC Code:
12000000
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
solid phase microextraction (SPME): suitable
application(s)
food and beverages
compatibility
for use with Non-CTC CombiPAL Type Autosamplers
greener alternative category
General description
The holder protects the coated fiber, and controls exposure of the fiber during analyte adsorption and desorption. The holder is reusable indefinitely and accepts the replaceable fiber assembly. First time users must order both a holder and a fiber assembly.
Fiber Holder for Manual Sampling
An adjustable depth guide positions the fiber for sampling and for correct placement in the heated zone of the GC injection port. The fiber can be locked in the exposed position.
Fiber Holder for Automated Sampling or HPLC Analysis
Use this fiber holder with a Varian 8100/8200 AutoSampler or with our SPME/HPLC interface. An SPME upgrade kit is necessary for operation with the Varian AutoSampler - contact Varian Instrument Division for information concerning system requirements.
Fiber Holder for CTC CombiPAL/CombiPAL-xt and Gerstel MPS2
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.
Legal Information
CombiPAL is a trademark of CTC Analytics
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)
Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)
Articles
A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.
A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.
A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.
A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.