Key Documents
57364-U
Supelco
SPME sampling stand holder & rod assembly
greener alternative
for use with SPME Sampling Stand
About This Item
UNSPSC Code:
41115854
NACRES:
NB.21
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
Quality Level
sustainability
Greener Alternative Product
technique(s)
solid phase microextraction (SPME): suitable
application(s)
food and beverages
compatibility
for use with SPME Sampling Stand
greener alternative category
General description
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)