Skip to Content
Merck
All Photos(2)

Key Documents

57348-U

Supelco

SPME Fiber Assembly

greener alternative

DVB/CAR/PDMS, for use with manual holder or autosampler, Nitinol-core (NIT), 24 ga, 50 μm (DVB layer), 30 μm (CAR/PDMS layer)

Synonym(s):

StableFlex 2CM SPME FIBER PK3,50/30UM DVB/CARBOXEN-PDMS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

Product Name

SPME Fiber Assembly, needle size 24 ga, matrix active group 50/30µm DVB/CAR/PDMS, StableFlex fiber, for use with autosampler, fiber L 2 cm, pk of 3

material

StableFlex fiber
notched gray hub

Quality Level

needle size

24 ga

description

StableFlex

packaging

pk of 3

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

30 μm (CAR/PDMS layer)
50 μm (DVB layer)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

2 cm

matrix active group

50/30µm DVB/CAR/PDMS
DVB/CAR/PDMS coating

application(s)

food and beverages

compatibility

for analyte group flavors (volatiles and semivolatiles, C3-20) (MW 40-275)
for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

Odor Compounds

Other Notes

Appearance may vary. Darker spots may be visible on the fiber but will not affect performance.

Footnote

Coating bonded to a flexible fused silica core, yielding a more suitable coating on a less breakable fiber. There may be a slight difference in extraction selectivity compared to the same coating on a standard fused silica core. 2cm fiber assembly contains no spring.

Legal Information

StableFlex is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Devin L Maurer et al.
Scientific reports, 9(1), 12103-12103 (2019-08-23)
The primary hurdle for diagnosis of some diseases is the long incubation required to culture and confirm the presence of bacteria. The concept of using microbial VOCs as "signature markers" could provide a faster and noninvasive diagnosis. Finding biomarkers is
M Caldeira et al.
Journal of chromatography. A, 1254, 87-97 (2012-07-28)
Allergic asthma represents an important public health issue, most common in the paediatric population, characterized by airway inflammation that may lead to changes in volatiles secreted via the lungs. Thus, exhaled breath has potential to be a matrix with relevant
Nobuo Ochiai et al.
Journal of chromatography. A, 1240, 59-68 (2012-05-01)
A method for analysis of a wide range of odor compounds in aqueous samples at sub-ng mL⁻¹ to μg mL⁻¹ levels was developed by full evaporation dynamic headspace (FEDHS) and gas chromatography-mass spectrometry (GC-MS). Compared to conventional DHS and headspace
Inés Racamonde et al.
Journal of chromatography. A, 1245, 167-174 (2012-06-05)
A new method for the determination of two cannabinoids, Δ9-tetrahydrocannabinol (THC) and 11-nor-9-carboxy-Δ9-tetrahydrocannabinol (THCCOOH), in wastewater samples is proposed using solid-phase microextraction (SPME) as extraction technique and gas chromatography coupled to mass spectrometry (GC-MS) as determination technique. Several parameters involved
Amparo Gamero et al.
Journal of chromatography. A, 1272, 1-7 (2012-12-19)
MicroVinification platforms are used for screening purposes to study aroma development in wine. These high-throughput methodologies require flavor analysis techniques that allow fast detection of a high number of aroma compounds which often appear in very low concentrations (μg/l). In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service