Skip to Content
Merck
All Photos(3)

Key Documents

57439-U

Supelco

SPME Fiber Assembly

greener alternative

PDMS/DVB, Overcoated, for use with autosampler or manual holder, Nitinol-core (NIT), 23 ga, 75 μm (Coating thickness includes 65 μm + 10 μm OC (overcoating))

Synonym(s):

SPME-Overcoated Fiber Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

Product Name

SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm(Coating thickness includes 65 μm coating + 10 μm OC (overcoating), needle size 23 ga, pkg of 3 ea, fiber L 1 cm

material

StableFlex fiber (Polydimethylsiloxane/Divinylbenzene (PDMS/DVB))
notched pink hub

Quality Level

needle size

23 ga

description

Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) fiber overcoated with a layer of PDMS.

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating))

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS/DVB coating, Overcoated Polydimethylsiloxane/Divinylbenzene

application(s)

food and beverages

compatibility

for use with autosampler or manual

greener alternative category

General description

For direct immersion into matrix or headspace analysis of volatile or semi-volatile analytes (MW 50-300+)
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application


  • A solid phase microextraction Arrow with zirconium metal-organic framework/molybdenum disulfide coating coupled with gas chromatography-mass spectrometer for the determination of polycyclic aromatic hydrocarbons in fish samples: This study highlights the effectiveness of PDMS/DVB SPME fibers in detecting polycyclic aromatic hydrocarbons in fish, showcasing their potential in environmental and food safety applications (Yuan Y, Lin X, Li T, 2019).

  • Applicability of the new 60 μm polyethylene glycol solid-phase microextraction fiber assembly for the simultaneous analysis of six pesticides in water: This research illustrates the use of SPME fibers for the efficient and simultaneous analysis of multiple pesticides in water samples, underscoring their value in environmental monitoring and chemical analysis (Pereira A, Silva E, Cerejeira MJ, 2014).

  • Passive sampling of ambient ozone by solid phase microextraction with on-fiber derivatization: This study demonstrates the utility of SPME fibers for passive air sampling and analysis of ozone, providing a valuable tool for environmental monitoring and analysis of atmospheric compounds (Lee IS, Tsai SW, 2008).

  • Diffusive sampling of airborne furfural by solid-phase microextraction device with on-fiber derivatization: This publication showcases the application of SPME fibers for capturing and analyzing airborne furfural, highlighting their use in air quality monitoring and industrial environments (Tsai SW, Kao KY, 2006).


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther De La Fuente et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 44(10), 1004-1010 (2009-10-16)
The complex molecular assemblages were analysed in the soil gas phase after applying pine forest wastes (PFW) or sugarbeet vinasses (SBV) for soil-borne crop pests' management. For this purpose, solid-phase micro-extraction (SPME) and gas chromatography (GC) were used coupled with
Marghereth Saverini et al.
Journal of photochemistry and photobiology. B, Biology, 108, 8-15 (2012-01-20)
The presence of (±)α-pinene, (+)β-pinene, (+)3-carene, and R-(+)limonene terpenes in wastewater of a citrus transformation factory was detected and analyzed, in a previous study, by using Solid Phase Micro-extraction (SPME) followed by GC analyses. Purpose of that research was to
Y Bezman et al.
Journal of agricultural and food chemistry, 49(11), 5425-5432 (2001-11-21)
The occurrence of methional in fresh orange juice, and possible occurrence of beta-damascenone in heated orange juice, has been previously suggested. Here we report on the occurrence of 2-methyl-3-furanthiol in the headspace, collected by solid-phase micro-extraction, of fresh, pasteurized, and
M A Stack et al.
Chemosphere, 41(11), 1821-1826 (2000-11-01)
Solid phase micro extraction (SPME) was applied to the determination of selected trihalomethanes (THMs), chloroform, bromodichloromethane, dibromochloromethane, bromoform, in potable and recreational waters. The selected samples were environmentally significant due to mandatory limits imposed by regulatory agencies. Extraction of the
Mona Syhre et al.
Tuberculosis (Edinburgh, Scotland), 88(4), 317-323 (2008-02-26)
Worldwide, tuberculosis (TB) kills nearly 2 million people annually, yet rapid diagnosis still relies on a 100-year-old method of sputum staining for acid-fast bacilli. The advent of solid phase micro-extraction and gas chromatography/mass spectrometry makes it possible to systematically investigate

Articles

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

A direct immersion SPME method using a new overcoated SPME fiber was developed for the extraction of pesticides from pureed prune baby food.

See All

Protocols

Overcoated SPME fiber enables low-level BPA analysis in food via SPME-GC-MS/MS, meeting tighter migration limit regulations.

Overcoated SPME fiber enables low-level BPA analysis in food via SPME-GC-MS/MS, meeting tighter migration limit regulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service