Skip to Content
Merck
All Photos(1)

Key Documents

I0408

Sigma-Aldrich

Invertase Glycoprotein Standard

BioReagent, from Saccharomyces cerevisiae, for proteomics

Synonym(s):

Invertase from baker’s yeast (S. cerevisiae), β-D-Fructofuranosidase, β-D-Fructofuranoside fructohydrolase, Saccharase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Saccharomyces cerevisiae

Quality Level

grade

for proteomics

product line

BioReagent

form

lyophilized powder

mol wt

60 kDa

concentration

≥0.5 mg/vial protein (E1%/280)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Invertase Glycoprotein Standard has been used:
  • to generate N-linked glycan library
  • as a negative control to study the binding of lectins to high mannose structures
  • for sample pre-treatment in proteomic analyses to study drug-induced toxic epidermal necrolysis
The Invertase Glycoprotein Standard can be used to demonstrate N-glycosylation using PNGase F with both in-solution and in-gel procedures. The extent of deglycosylation can be assessed by mobility shift on SDS-PAGE gels.
Used in the production of confectionary foods and artificial honey.

Biochem/physiol Actions

Invertase hydrolyzes sucrose into glucose and fructose yielding a colorless product, unlike acid hydrolysis which produces colored products.

Other Notes

Invertase is an enzyme that catalyses the hydrolysis of sucrose into fructose and glucose. Invertase Glycoprotein Standard is the periplasmic (glycosylated form, external invertase) with 50% of its mass as polymannan. Since yeast can provide an alternative system for protein glycosylation that is similar to mammalian systems, periplasmic invertase is often used as a model for the study of the function of oligosaccharides in glycoproteins and for studies on glycoprotein biosynthesis.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteomic kinetic analysis of blister fluid and serum in a patient with drug-induced toxic epidermal necrolysis. A comparison with skin immunohistochemistry
Paquet P, et al.
Current Drug Safety (2012)
Glycan characterization of the NIST RM monoclonal antibody using a total analytical solution: From sample preparation to data analysis
Hilliard M, et al.
MAbs (2017)
A lectin affinity workflow targeting glycosite-specific, cancer-related carbohydrate structures in trypsin-digested human plasma
Analytical biochemistry (2011)
M Angela Sainz-Polo et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 68(Pt 12), 1538-1541 (2012-11-30)
Saccharomyces cerevisiae invertase (ScInv) is an enzyme encoded by the SUC2 gene that releases β-fructose from the nonreducing termini of various β-D-fructofuranoside substrates. Its ability to produce 6-kestose by transglycosylation makes this enzyme an interesting research target for applications in
Mei-Liang Zhou et al.
Glycobiology, 22(12), 1775-1785 (2012-08-11)
The raffinose family oligosaccharides (RFOs), such as raffinose and stachyose, are synthesized by a set of distinct galactosyltransferases, which sequentially add galactose units to sucrose. The accumulation of RFOs in plant cells are closely associated with the responses to environmental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service