Skip to Content
Merck
All Photos(1)

Key Documents

D8144

Sigma-Aldrich

Dextranase from Penicillium sp.

lyophilized powder, 400-800 units/mg protein

Synonym(s):

1,6-α-D-Glucan 6-glucanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Penicillium sp.

Quality Level

form

lyophilized powder

specific activity

400-800 units/mg protein

composition

Protein, 35% Lowry

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dextranase from fungi belong to the glycoside hydrolase families (GH) 49. Dextranase from Penicillium sp. are stable at varying temperature and pH.

Application

Dextranase from Penicillium sp has been used:
  • in the digestion of exopolysaccharides from P. aeruginosa biofilms and dextran polymers
  • as a component of trypticase soy broth (TSYPS) for Streptococcus mutans culture
  • in microgel degradation

Biochem/physiol Actions

An endodextranase that hydrolyzes -(1,6)-glucosidic linkages in dextran. Dextrans are undesirable compounds synthesized from sucrose by microbial contaminants during sugar production that increase viscosity of the flow and decrease industrial recovery. Dextranase has been used for hydrolyzing dextran at sugar mills in order to improve efficiency of sugar production.

Unit Definition

One unit will liberate 1.0 μmole of isomaltose (measured as maltose) per min at pH 6.0 at 37 °C, using dextran as substrate.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dextranase from Penicillium minioluteum: reaction course, crystal structure, and product complex
Larsson AM, et al.
Structure, 11(9), 1111-1121 (2003)
Purification & properties of an extracellular dextranase from Penicillium janthinellum.
B Dasmahapatra et al.
Indian journal of biochemistry & biophysics, 15(3), 206-210 (1978-06-01)
Hordenine: a novel quorum sensing inhibitor and antibiofilm agent against Pseudomonas aeruginosa
Zhou JW, et al.
Journal of Agricultural and Food Chemistry, 66(7), 1620-1628 (2018)
Biology and genome sequence of Streptococcus mutans phage M102AD
Delisle AL, et al.
Applied and Environmental Microbiology, 78(7), 2264-2271 (2012)
Label-free high-throughput functional lytic assays
O'Malley SM, et al.
Journal of Biomolecular Screening, 12(1), 117-125 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service