Skip to Content
Merck
All Photos(1)

Key Documents

I9274

Supelco

Invertase from baker′s yeast (S. cerevisiae)

200-300 units/mg solid

Synonym(s):

β-D-Fructofuranosidase, β-D-Fructofuranoside fructohydrolase, Saccharase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21

Quality Level

specific activity

200-300 units/mg solid

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

foreign activity

α-glucosidase (maltase), phosphoglucomutase, phosphoglucose isomerase ≤0.01%
6-phosphogluconic dehydrogenase, α-galactosidase, β-galactosidase, and NADH oxidase ≤0.001%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Invertase is encoded by the SUC2 (Saccharomyces cerevisiae S288C) gene. In yeast this enzyme exist in: glycosylated (secreted) and non-glycosylated (intracellular) form. The external invertase is composed of carbohydrate (50%) and 60,000 D subunits.

Application

Invertase from baker′s yeast (S. cerevisiae) has been used:
  • in liquid media (YES) and is used to determine the invertase activity
  • supplemented to the reaction mixture in the extraction and analysis of sugars and starch
  • for sucrose hydrolysis

Used in the production of confectionary foods and artificial honey.

Biochem/physiol Actions

Invertase hydrolyzes sucrose into glucose and fructose yielding a colorless product, unlike acid hydrolysis which produces colored products.

Unit Definition

One unit will hydrolyze 1.0 μmole sucrose to glucose and fructose per minute at pH 4.6 at 25 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subunit structure of external invertase from Saccharomyces cerevisiae.
Trimble RB and Maley F
The Journal of Biological Chemistry, 252(12), 4409-4412 (1977)
Two differentially regulated mRNAs with different 5? ends encode secreted and intracellular forms of yeast invertase
Carlson M and Botstein D
Cell, 28(1), 145-154 (1982)
Production of butyric acid by Clostridium tyrobutyricum (ATCC25755) using sweet sorghum stalks and beet molasses
Sjoblom M, et al.
Industrial Crops and Products, 74, 535-544 (2015)
Carbohydrate profiles during cotton floral bud (square) development
Tarpley L and Sassenrath GF
Journal of Agromedicine, 192(5), 363-372 (2006)
Morphological and physiological changes in Streptomyces lividans induced by different yeasts
Santamaria R, et al.
Archives of Microbiology, 177(3), 259-266 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service