- Crystallization and preliminary X-ray diffraction analysis of the invertase from Saccharomyces cerevisiae.
Crystallization and preliminary X-ray diffraction analysis of the invertase from Saccharomyces cerevisiae.
Acta crystallographica. Section F, Structural biology and crystallization communications (2012-11-30)
M Angela Sainz-Polo, Alvaro Lafraya, Aitana Polo, Julia Marín-Navarro, Julio Polaina, Julia Sanz-Aparicio
PMID23192042
ABSTRACT
Saccharomyces cerevisiae invertase (ScInv) is an enzyme encoded by the SUC2 gene that releases β-fructose from the nonreducing termini of various β-D-fructofuranoside substrates. Its ability to produce 6-kestose by transglycosylation makes this enzyme an interesting research target for applications in industrial biotechnology. The native enzyme, which presents a high degree of oligomerization, was crystallized by vapour-diffusion methods. The crystals belonged to space group P3(1)21, with unit-cell parameters a=268.6, b=268.6, c=224.4 Å. The crystals diffracted to 3.3 Å resolution and gave complete data sets using a synchrotron X-ray source.