Skip to Content
Merck
All Photos(1)

Key Documents

G2174

Sigma-Aldrich

β-Glucuronidase from limpets (Patella vulgata)

aqueous solution, ≥85,000 units/mL

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

limpet (Patella vulgata)

Quality Level

form

aqueous solution

specific activity

≥85,000 units/mL

secondary activity

≤7,500 units/mL sulfatase

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

New Technical Article Comparing Performance of Different Enzymes
Learn more about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine
The enzyme from patella vulgata is reported to be more effective in hydrolyzing opioid-glucuronides than the Helix pomatia, bovine liver and E. coli enzymes

Features and Benefits

Save time with no reconstitution in this convenient liquid format.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 3.8 (30 min assay).
Sulfatase Unit Definition: One unit of sulfatase will hydrolyze 1.0 μmole p-nitrocatechol sulfate per hr at pH 5.0 at 37 °C.

Physical form

Aqueous solution in 0.9% NaCl with 0.02% sodium azide as preservative.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Combie et al.
Clinical chemistry, 28(1), 83-86 (1982-01-01)
beta-Glucuronidase from Patella vulgata, Helix aspersa, Helix pomatia, and bovine liver were evaluated for usefulness in routine hydrolysis of drug-glucuronic acid conjugates from equine urine samples. Factors affecting the reaction rate (enzyme concentration, ligand concentration, temperature, and pH) were optimized.
Vanessa Andrea Meling et al.
Molecules (Basel, Switzerland), 27(8) (2022-04-24)
The aquaculture industry has become a sustainable source of food for humans. Remaining challenges include disease issues and ethical concerns for the discomfort and stress of farmed fish. There is a need for reliable biomarkers to monitor welfare in fish
T A Jennison et al.
Journal of analytical toxicology, 17(4), 208-210 (1993-07-01)
Methods to confirm morphine in urine require hydrolysis to liberate morphine from its 3-beta-D glucuronide (M-3G) conjugate. Lengthy enzyme hydrolysis procedures prolong testing turnaround time whereas rapid enzyme methods may produce a low conversion of M-3G to morphine. The purpose
Miribane Dërmaku-Sopjani et al.
Molecular membrane biology, 30(8), 369-385 (2013-10-16)
The Klotho gene was identified as an 'aging suppressor' in mice. Overexpression of the Klotho gene extends lifespan and defective Klotho results in rapid aging and early death. Both the membrane and secreted forms of Klotho have biological activity that
Akihiro Imura
Clinical calcium, 23(10), 1476-1482 (2013-10-01)
Recent understandings of phosphate regulation have, at least in part, depended upon the findings of fibroblast growth factor (FGF) 23, the hormone specific for phosphate regulation. Moreover, FGF23 would be the most important marker for prognosis in chronic kidney disease

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

See All

Chromatograms

application for SPE, application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service