Skip to Content
Merck
All Photos(9)

Key Documents

G8132

Sigma-Aldrich

β-Glucuronidase from limpets (Patella vulgata)

Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

limpet (Patella vulgata)

Quality Level

type

Type L-II

form

lyophilized powder

specific activity

1,000,000-3,000,000 units/g solid

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

β-Glucuronidase Type L-II has been shown to be a superior enzyme for the hydrolysis of drug-glucuronides from urine.
New Technical Article Comparing Performance of Different Enzymes
Learn more about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine
The enzyme from patella vulgata is reported to be more effective in hydrolyzing opioid-glucuronides than the Helix pomatia, bovine liver and E. coli enzymes

Biochem/physiol Actions

β-glucuronidase (β-GIc) is an exoglycosidase that catalyzes the breakdown of complex carbohydrates. In humans it converts conjugated bilirubin into the unconjugated form, making bilirubin suitable for reabsorption. Morphine 3-β-D glucuronide was converted to morphine using ß-glucuronidase obtained from Patella vulgate.

Quality

Contains sulfatase activity which is inhibited by 0.1 M phosphate.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 3.8 (30 min assay).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vectors with the gus reporter gene for identifying and quantitating promoter regions in <I>Saccharomyces cerevisiae</I>
S.V. Marathe &amp; J.E. McEwen
Gene, 154, 105-107 (1994)
Robert West et al.
Pain physician, 13(1), 71-78 (2010-02-02)
Physicians determine patient compliance with their medications by use of urine drug testing. It is known that measurement of benzodiazepines is limited by immunoassay specificity and cutoff limits and therefore does not offer physicians an accurate picture of their patients'
Catalytic mechanisms of enymatic glycosyl transfer
M.L. Sinnott
Chemical Reviews, 90, 1171-1202 (1990)
J Combie et al.
Clinical chemistry, 28(1), 83-86 (1982-01-01)
beta-Glucuronidase from Patella vulgata, Helix aspersa, Helix pomatia, and bovine liver were evaluated for usefulness in routine hydrolysis of drug-glucuronic acid conjugates from equine urine samples. Factors affecting the reaction rate (enzyme concentration, ligand concentration, temperature, and pH) were optimized.
S Feng et al.
Journal of analytical toxicology, 25(7), 589-593 (2001-10-16)
Buprenorphine, which is a powerful analgesic, a substitution drug for opioids widely used in Europe, and a promising new drug currently undergoing clinical trials in the treatment of opioid dependence in the U.S., is excreted in human urine mainly as

Articles

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

See All

Protocols

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service