Skip to Content
Merck
All Photos(2)

Key Documents

G8420

Sigma-Aldrich

β-Glucuronidase from Escherichia coli

≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli overproducing strain

Quality Level

form

lyophilized powder

specific activity

≥20,000 units/mg protein

mol wt

290 kDa

composition

, ~25%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine
Effective in the hydrolysis of steroid glucuronides.
Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Quality

Contains buffer salts and stabilizer.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at the pH 6.8 (30 min assay).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymes
Hans Ulrich Bergmeyer
Methods of Analysis, 2, 460-461 (1974)
V Graef et al.
Clinical chemistry, 23(3), 532-535 (1977-03-01)
We determined the enzymic activity of beta-glucuronidase preparations from bovine liver, Helix pomatia, and Escherischia coli with steroid glucuronides and nonsteroid glucuronides as substrates. We also studied the effect of Na2SO4 on the enzymic hydrolysis of several substrates with the
Fei Song et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 67(11-12), 611-619 (2013-02-19)
Genes coding for avenin-like proteins (ALP) represent a new family of wheat storage protein genes. To find a wheat endosperm-specific promoter, a 1644-bp fragment upstream of the ALP type-B gene (GenBank accession number JN622144) was isolated. The important promoter elements
Regina Goetz et al.
Nature reviews. Molecular cell biology, 14(3), 166-180 (2013-02-14)
Fibroblast growth factors (FGFs) mediate a broad range of functions in both the developing and adult organism. The accumulated wealth of structural information on the FGF signalling pathway has begun to unveil the underlying molecular mechanisms that modulate this system
Xiaoyan Liu et al.
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences, 42(1), 67-74 (2013-03-19)
To construct a lentiviral RNA interference system targeting heparanase (HPSE) based on miR30 and to test its silencing effect. Three heparanase-shRNA structures were designed based miR30. The targeting fragments were obtained by PCR, then inserted into the vector LV PP-GFP

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

See All

Protocols

Objective: To standardize a procedure for the enzymatic assay of β-Glucuronidase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service