Skip to Content
Merck
All Photos(8)

Key Documents

G7017

Sigma-Aldrich

β-Glucuronidase from Helix pomatia

Type HP-2, aqueous solution, ≥100,000 units/mL

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Helix pomatia

Quality Level

type

Type HP-2

form

aqueous solution

specific activity

≥100,000 units/mL

secondary activity

≤7,500 units/mL sulfatase

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Glucuronidase Type H-2 from Helix pomatia is a crude solution of enzymes derived from the digestive juices of the Roman snail. β-glucuronidases derived from mollusks often contain sulfatase activity. Both activities are useful in the hydrolysis of metabolites for toxicology and biomarker analysis. For a broader look at our β-Glucuronidase product line and their uses see our page about Drug Metabolite Analysis

Application

  • Sample preparation for LC/MS through digestion of glucuronide conjugated compounds for biomarker and toxicology research.
  • Removal of glucuronide conjugates such as for the preparation of Baicalein.­
New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine

Biochem/physiol Actions

Endogenous beta-glucuronidase is a lysosomal enzyme involved in digestion of glucuronide-modified catabolites.
Microbial beta-glucuronidase found in microbiota can cause re-absorption of metabolites by hydrolysis of glucuronide conjugates of xenobiotic and endobiotic metabolites.
Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Features and Benefits

  • pH optima 4.0 - 5.0.
  • Sulfatase pH optima 6.2
  • Low Sulfatase activity (<7,500 units/mL).
  • Ammonium Sulfate suspension.
  • Mollusk-derived beta-glucuronidase.
  • Useful for acidic hydrolysis of glucuronides in low pH samples.

Quality

Many β-glucuronidases derived from mollusks also contain sulfatase activity.

Linkage

Similar to G0876, but produced by Sigma.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 5.0 (30 min assay).
Sulfatase Unit Definition: One unit of sulfatase will hydrolyze 1.0 μmole p-nitrocatechol sulfate per hr at pH 5.0 at 37 °C.

Physical form

Aqueous solution in ~1.0 M ammonium sulfate with 3 mM sodium azide as preservative.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Bláhová et al.
Journal of dairy science, 99(9), 6973-6982 (2016-07-11)
Isoflavones are natural phytoestrogens with antioxidant and endocrine-disrupting potencies. Monitoring of their levels is important to ensure the high quality and safety of food, milk, and dairy products. The efficiency and accuracy of phytoestrogen analyses in complex matrices such as
Mladen Boban et al.
Journal of cardiovascular pharmacology, 47(5), 695-701 (2006-06-16)
By using red wine (RW), dealcoholized red wine (DARW), polyphenols-stripped red wine (PSRW), ethanol-water solution (ET), and water (W), the role of wine polyphenols, ethanol, and urate on vascular function was examined in humans (n = 9 per beverage) and
W Dennis Ulrey et al.
Journal of AOAC International, 96(4), 917-924 (2013-09-05)
A multilaboratory study of AOAC Official Method 2011.23 was performed to satisfy requirements for Final Action status through the AOAC expert review panel process. The study included nine collaborating laboratories from the United States, Canada, Brazil, and The Netherlands. Five
Monica L Vermillion Maier et al.
Drug metabolism and disposition: the biological fate of chemicals, 49(8), 694-705 (2021-05-27)
3,3'-Diindolylmethane (DIM), a major phytochemical derived from ingestion of cruciferous vegetables, is also a dietary supplement. In preclinical models, DIM is an effective cancer chemopreventive agent and has been studied in a number of clinical trials. Previous pharmacokinetic studies in
Madalina Neacsu et al.
Molecular nutrition & food research, 61(3) (2016-07-01)
Phytophenols present in cereals are metabolised to compounds that could be partly responsible for the reduced risk of chronic diseases and all-cause mortality associated with fibre-rich diets. The bioavailability, form and in vivo concentrations of these metabolites require to be

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Protocols

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service