Skip to Content
Merck
All Photos(1)

Key Documents

PHR1527

Supelco

Dexamethasone Acetate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dexamethasone 21-acetate, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione-21-acetate, 9α-Fluoro-16α-methylprednisolone-21-acetate, Prednisolone F acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
Beilstein:
2342608
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

Quality Level

grade

certified reference material
pharmaceutical secondary standard

Agency

BP
EP
USP
traceable to Ph. Eur. D0710000
traceable to USP 1176506

vapor pressure

<0.0000001 kPa ( 25 °C)

API family

dexamethasone

CofA

current certificate can be downloaded

packaging

pkg of 1 g

storage condition

protect from light (20 mm aluminium crimp seal for unused portion)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

460.4-464 °F (238—240°C)

solubility

acetone: freely soluble
dichloromethane: slightly soluble
methanol: freely soluble
water: practically insoluble

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-30°C

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@@H](C)[C@]4(O)C(=O)COC(C)=O

InChI

1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1

InChI key

AKUJBENLRBOFTD-RPRRAYFGSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Dexamethasone acetate (Decadron-LA) belongs to the class of long-acting glucocorticoids. Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Dexamethasone Acetate is a steroid used for the treatment of various diseases because of its anti-inflammatory and inmunosuppresor effects. It is a common component of ointments, creams, lotions, aerosols and microemulsions.

Application

Dexamethasone Acetate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.
Dexamethasone acetate is used as a pharmaceutical reference standard for the quantification the analyte in pharmaceutical formulations using liquid chromatography (LC) technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3645 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of HPLC method for analysis of dexamethasone acetate in microemulsions.
Urban MCC, et al.
Brazilian Journal of Pharmaceutical Sciences , 45(1), 87-92 (2009)
Daniele de Paula Faria et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(8), 1330-1335 (2014-06-11)
The experimental autoimmune encephalomyelitis model is a model of multiple sclerosis that closely mimics the disease characteristics in humans. The main hallmarks of multiple sclerosis are neuroinflammation (microglia activation, monocyte invasion, and T-cell infiltration) and demyelination. PET imaging may be
Determination of dexamethasone acetate in cream by HPLC.
Garcia CV, et al.
Journal of Pharmaceutical and Biomedical Analysis, 31(3), 597-600 (2003)
Dexamethasone
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 10(3), 4961-4963 (2021)
Dunja Drakulić et al.
Journal of molecular neuroscience : MN, 55(4), 959-967 (2014-11-05)
Although dexamethasone (DEX), a synthetic glucocorticoid receptor (GR) analog with profound effects on energy metabolism, immune system, and hypothalamic-pituitary-adrenal axis, is widely used therapeutically, its impact on the brain is poorly understood. The aim of the present study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service