Skip to Content
Merck
All Photos(1)

Key Documents

Y0001593

Dexamethasone for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dexamethasone, (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
Beilstein:
2066651
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

dexamethasone

form

powder

manufacturer/tradename

EDQM

storage condition

protect from light

mp

262-264 °C (lit.)
262-264 °C

solubility

water: <0.1 g/L

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Dexamethasone belongs to the class of synthetic glucocorticoid steroid drugs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone is used as an EP referencs standard to quantify the analyte in pharmaceutical formulations using liquid chromatography (LC) technique.

Biochem/physiol Actions

Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dexamethasone: Therapeutic Uses, Mechanism of Action and Potential Side Effects (2013)
Dexamethasone
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 10(3), 4961-4963 (2021)
Sara Bringhen et al.
Blood, 124(1), 63-69 (2014-05-24)
This multicenter, open-label phase 2 trial determined the safety and efficacy of carfilzomib, a novel and irreversible proteasome inhibitor, in combination with cyclophosphamide and dexamethasone (CCyd) in patients with newly diagnosed multiple myeloma (NDMM) ≥65 years of age or who
Lotfi Benboubker et al.
The New England journal of medicine, 371(10), 906-917 (2014-09-04)
The combination melphalan-prednisone-thalidomide (MPT) is considered a standard therapy for patients with myeloma who are ineligible for stem-cell transplantation. However, emerging data on the use of lenalidomide and low-dose dexamethasone warrant a prospective comparison of the two approaches. We randomly
Wei Zheng et al.
Genes & development, 28(14), 1592-1603 (2014-07-18)
Primitive lymphatic vessels are remodeled into functionally specialized initial and collecting lymphatics during development. Lymphatic endothelial cell (LEC) junctions in initial lymphatics transform from a zipper-like to a button-like pattern during collecting vessel development, but what regulates this process is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service