
Dexamethasone acetate

Synonym(s):
Dexamethasone 21-acetate, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione-21-acetate, 9α-Fluoro-16α-methylprednisolone-21-acetate, Prednisolone F acetate
Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
Products
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3