Skip to Content
Merck
All Photos(1)

Key Documents

D0718000

Dexamethasone pivalate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dexamethasone 21-trimethylacetate, 9-Fluoro-11β,17-dihydroxy-16α-methyl-3,20-dioxopregna-1,4-dien-21-yl 2,2-dimethylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dexamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

F[C@@]21[C@H]([C@H]4[C@@]([C@@]([C@@H](C4)C)(O)C(=O)COC(=O)C(C)(C)C)(C[C@@H]2O)C)CCC3=CC(=O)C=C[C@@]31C

InChI

1S/C27H37FO6/c1-15-11-19-18-8-7-16-12-17(29)9-10-24(16,5)26(18,28)20(30)13-25(19,6)27(15,33)21(31)14-34-22(32)23(2,3)4/h9-10,12,15,18-20,30,33H,7-8,11,13-14H2,1-6H3/t15-,18+,19+,20+,24+,25+,26+,27+/m1/s1

InChI key

XMQRFWOHEPJZPS-JHUFMRCFSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone pivalate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of glucocorticoids and prematurity on absorption of colostral immunoglobulin in the calf.
N E Johnston et al.
Australian veterinary journal, 63(6), 191-192 (1986-06-01)
R J Fairclough et al.
Australian journal of biological sciences, 34(3), 313-319 (1981-01-01)
A radioimmunoassay has been developed for the measurement of dexamethasone in plasma and milk of cows injected with long- and short-acting dexamethasone esters. Dexamethasone antiserum was produced by injecting cows with a dexamethasone-21-hemisuccinate-human serum albumin complex. The antisera was highly
P D Mansell et al.
Australian veterinary journal, 84(9), 312-316 (2006-09-09)
To determine the effect of induction of parturition on health, milk production and reproductive performance of dairy cows. A prospective cohort study in 62 dairy herds. Health, milk production and fertility indices were documented for 1449 dairy cows treated with
Betamethasone dipropionate (0.05%) plus salicylic acid (3%) ointment versus dexamethasone trimethyl acetate (1%) and salicylic acid 4% ointments in chronic dermatoses.
S P Chattopadhyay et al.
Indian journal of dermatology, 32(2), 41-44 (1987-04-01)
S Lloyd et al.
Zeitschrift fur Parasitenkunde (Berlin, Germany), 66(1), 57-61 (1981-01-01)
Three, six-month-old dogs were infected with 2,000 eggs of Toxocara canis. Three weeks after infection 15 immature adult parasites were found in the intestine of the dog which had been treated with high doses of corticosteroids (2 ml of Opticortenol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service