Skip to Content
Merck
All Photos(3)

Key Documents

M7252

Sigma-Aldrich

17α-Methyltestosterone

solid (photosensitive)

Synonym(s):

17α-Methyl-4-androsten-17β-ol-3-one, 17β-Hydroxy-17α-methyl-4-androsten-3-one, Mesterone, Methyltestosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Beilstein:
2057425
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

solid (photosensitive)

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

mp

162-168 °C (lit.)

solubility

H2O: ≤0.5 mg/mL
ethanol: 22 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 4.8 mg/mL

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C20H30O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h12,15-17,22H,4-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1

InChI key

GCKMFJBGXUYNAG-HLXURNFRSA-N

Gene Information

human ... AR(367)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Methyltestosterone (Mesterone) is an androgen and anabolic steroid.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Herfeld et al.
Hormones and behavior, 34(3), 303-319 (1999-01-08)
Adult males of African weakly discharging electric fish (family: Mormyridae) are distinguished from juveniles and adult females by a dorsally directed indentation of the posterior ventral body wall and by massive bone expansion of the bases of a select number
C Gómez et al.
Steroids, 78(1), 44-52 (2012-11-07)
Methyltestosterone (MT) is one of the most frequently detected anabolic androgenic steroids in doping control analysis. MT misuse is commonly detected by the identification of its two main metabolites excreted as glucuronide conjugates, 17α-methyl-5α-androstan-3α,17β-diol and 17α-methyl-5β-androstan-3α,17β-diol. The detection of these
Anne S Mortensen et al.
Chemosphere, 83(8), 1035-1044 (2011-03-01)
In the present study, groups of juvenile Atlantic salmon (Salmo salar) were fed gelatine capsules containing fish-food spiked with PFOA or PFOS (0.2 mg kg(-1) fish) and solvent (methanol). The capsules were given at days 0, 3 and 6. Blood
Carlos A A Penatti et al.
Neuropharmacology, 61(4), 653-664 (2011-06-08)
Disruption of reproductive function is a hallmark of abuse of anabolic androgenic steroids (AAS) in female subjects. To understand the central actions of AAS, patch clamp recordings were made in estrous, diestrous and AAS-treated mice from gonadotropin releasing hormone (GnRH)
Jingzhu Wang et al.
Analytical and bioanalytical chemistry, 406(24), 6061-6067 (2014-08-12)
Progesterone (PROG) is a naturally occurring progestagen, which has been used to prevent preterm birth, control persistent anovulatory bleeding, and treat premenstrual syndrome in clinical practices. Studies on the metabolism of PROG have demonstrated that PROG is the precursor of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service