Skip to Content
Merck
All Photos(1)

Key Documents

8.08913

Sigma-Aldrich

Zirconium(IV) chloride

anhydrous, for synthesis

Synonym(s):

Zirconium(IV) chloride, Zirconium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl4Zr
CAS Number:
Molecular Weight:
233.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-058-2
NACRES:
NA.22

vapor pressure

1.3 hPa ( 190 °C)

Quality Level

form

crystals

potency

1688 mg/kg LD50, oral (Rat)

pH

<1 (20 °C in H2O, saturated aqueous solution)

mp

437 °C

density

2.80 g/cm3 at 15 °C

bulk density

1200 kg/m3

storage temp.

2-30°C

InChI

1S/4ClH.Zr/h4*1H;/q;;;;+4/p-4

InChI key

DUNKXUFBGCUVQW-UHFFFAOYSA-J

General description

Zirconium(IV) chloride is a Lewis acid catalyst, widely used in organic synthesis for the preparation of useful key intermediates. Since it is a stable and environmentally friendly catalyst used in various chemical reactions such as carbon-carbon bond formation reactions, reduction reactions, protection, and deprotection reactions.

Application

Zirconium(IV) chloride can be used as a catalyst to synthesize:
  • β-amino alcohols by ring-opening of epoxides with aromatic amines.
  • β-amino analogs via conjugate addition of aliphatic amines to α,β-unsaturated ester, nitriles, and ketones.
  • 1,3,4-oxadiazole derivatives by cyclodehydration of 1,2-diacylhydrazines.
  • 3,4-Dihydropyrimidinone derivatives via Biginelli condensation reaction of aromatic aldehyde, β-ketoester, and urea.

Analysis Note

Assay (argentometric): ≥ 98.0 %
Hafnium (Hf): ≤ 40 ppm
Identity (ICP-OES): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zirconium (IV) Chloride Mediated Cyclodehydration of 1, 2-Diacylhydrazines: A Convenient Synthesis of 2, 5-Diaryl 1, 3, 4-Oxadiazoles
Sharma GVM, et al.
Synthetic Communications, 34(13), 2387-2391 (2004)
Zirconium (IV) chloride catalyzed ring opening of epoxides with aromatic amines
Swamy NR, et al.
Synthetic Communications, 34(4), 727-734 (2004)
Zirconium (IV) chloride catalyzed one-pot synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones
Reddy Ch Venkateshwar, et al.
Tetrahedron Letters, 43(14), 2657-2659 (2002)
Zirconium (IV) Chloride-Mediated Chemoselective Conjugate Addition of Aliphatic Amines to α, β-Ethylenic Compounds
Meshram HM, et al.
Synthetic Communications, 36(6), 795-801 (2006)
Applications of zirconium (IV) chloride in organic synthesis
Smitha G, et al.
Synthesis, 2008(06), 829-855 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service