Skip to Content
Merck
All Photos(1)

Key Documents

477028

Sigma-Aldrich

2-Hydroxyethyl methacrylate

≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate, HEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH2OH
CAS Number:
Molecular Weight:
130.14
Beilstein:
1071583
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

5 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 25 °C)

Assay

≥99%

contains

≤50 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.453 (lit.)

bp

67 °C/3.5 mmHg (lit.)

density

1.073 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCO

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxyethyl methacrylate (HEMA) is biocompatible in nature. Polymeric hydrogel scaffold can be produced by polymerizing HEMA in water.

Application

A polymeric hydrogel for drug delivery. Patent has been applied for the development of artificial cornea or KPro, it is composed of pHEMA (polymerized from HEMA) and poly (methyl methacrylate)(PMMA). HEMA may be used as a macromonomer for the synthesis of 2-hydroxyethylmethacrylate-poly(ε-caprolactone) (HEMA-PCL) by coordinated anionic ring opening polymerization (ROP). 

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiellini, F., et al.
Biomedical Polymers and Polymer Therapeutics, 63 (2001)
Synthesis of graft copolymers of poly(methacrylic acid)-g-poly(?-caprolactone) by coupling ROP and RAFT polymerizations
Kiehl J, et al.
Polymer, 53(3), 2012-2012 (2012)
Designing a gas foamed scaffold for keratoprosthesis
Zellander A, et al.
Materials Science and Engineering, C, 33(6), 3396-3403 (2013)
Hong Ying Li et al.
Journal of materials science. Materials in medicine, 24(8), 2001-2011 (2013-05-25)
Tissue engineered scaffolds and matrices have been investigated over the past decade for their potential in spinal cord repair. They provide a 3-D substrate that can be permissive for nerve regeneration yet have other roles including neuroprotection, altering the inflammatory
Duncan Chege et al.
Journal of acquired immune deficiency syndromes (1999), 65(5), 517-525 (2013-11-22)
To evaluate if systemic murine malarial infection enhances HIV susceptibility through parasite-induced mucosal immune alterations at sites of HIV sexual exposure. Malaria and HIV have a high degree of geographical overlap and interact substantially within coinfected individuals. We used a

Articles

With dentists placing nearly 100 million dental fillings into patients′ teeth annually in the U.S. alone, polymeric composite restoratives account for a very large share of the biomaterials market.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service