Skip to Content
Merck
All Photos(5)

Key Documents

221880

Sigma-Aldrich

Zirconium(IV) chloride

greener alternative

≥99.5% trace metals basis

Synonym(s):

Tetrachlorozirconium, Zirconium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrCl4
CAS Number:
Molecular Weight:
233.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.5% trace metals basis
form:
powder

vapor pressure

1 mmHg ( 190 °C)

Quality Level

Assay

≥99.5% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: zirconium

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤5000.0 ppm Trace Metal Analysis

transition temp

sublimation point 331 °C

density

2.8 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

Cl[Zr](Cl)(Cl)Cl

InChI

1S/4ClH.Zr/h4*1H;/q;;;;+4/p-4

InChI key

DUNKXUFBGCUVQW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

Direct amide formation from unactivated carboxylic acids and amines

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ines Sifaoui et al.
Frontiers in bioengineering and biotechnology, 9, 584115-584115 (2021-02-19)
In this study, the application of amphipods in vivo assays was evaluated. The main aim of this work was to check the potential use of this model in biocompatibility assessments of metal-organic frameworks (MOFs). Hence, six different MOFs were synthesized
Ilanit Doron-Mor et al.
Langmuir : the ACS journal of surfaces and colloids, 20(24), 10727-10733 (2004-11-17)
Coordination self-assembly of bishydroxamate-based metal-organic multilayers on gold employing a layer-by-layer (LbL) approach was investigated. It is shown that the solution chemistry of the participating metal ion has a marked influence on the composition and properties of the multilayers. Use
Stefania Fioravanti et al.
Organic & biomolecular chemistry, 10(41), 8207-8210 (2012-09-26)
ZrCl(4) was found to be an ideal catalyst to promote aza-Henry reactions between trifluoromethyl aldimines and some nitro alkanes giving new fluorinated β-nitro amines. The reaction is strongly influenced by the CF(3) group, the yield by the alkyl chain of
Harish Kumar et al.
Ultrasonics sonochemistry, 15(2), 129-132 (2007-04-17)
Sonication of aromatic aldehydes, urea and ethyl acetoacetate in presence of solvent (ethyl alcohol) or solvent-less dry media (bentonite clay) supported-zirconium (IV) chloride (ZrCl(4)) as catalyst at 35 kHz gives 6-methyl-4-substitutedphenyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid ethyl esters proficiently in high yields.
Giles W Theaker et al.
Dalton transactions (Cambridge, England : 2003), (48)(48), 6883-6885 (2008-12-04)
A group of readily available zirconium catalysts incapable of ethene-co-styrene polymerization are remarkably active and selective for the production of the new polymer ethene-co-tert-butylstyrene via a single site mechanism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service