Skip to Content
Merck
All Photos(3)

Key Documents

258202

Sigma-Aldrich

Hafnium(IV) chloride

98%

Synonym(s):

Hafnium tetrachloride, Tetrachlorohafnium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HfCl4
CAS Number:
Molecular Weight:
320.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 190 °C)

Quality Level

Assay

98%

form

powder

mp

432 °C (lit.)

solubility

H2O: decomposes(lit.)

SMILES string

Cl[Hf](Cl)(Cl)Cl

InChI

1S/4ClH.Hf/h4*1H;/q;;;;+4/p-4

InChI key

PDPJQWYGJJBYLF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hafnium(IV) chloride, also known as hafnium tetrachloride, is commonly used as a precursor in the production of hafnium metal and various hafnium compounds. Additionally, it serves as a catalyst in certain organic synthesis reactions. Due to its reactivity and sensitivity to moisture, hafnium(IV) chloride is typically handled under an inert atmosphere.

Application

Hafnium(IV) chloride can be used as:     
  • A precursor in the synthesis of lithium hafnium phosphate, which is a solid electrolyte material used in lithium batteries due to its high ionic conductivity and chemical stability.      
  • A catalyst in the acetalization process of various carbonyl compounds, including aldehydes and ketones.      
  • A catalyst in the direct ester condensation of carboxylic acids with alcohols.    
  • A high-capacity cathode material for lithium and sodium-ion batteries.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct condensation of carboxylic acids with alcohols catalyzed by hafnium (IV) salts.
Ishihara K, et al.
Science, 290.5494, 1140-1142 (2000)
Visualization of the cytoskeletal elements in tissue culture cells by bloc-staining with hafnium chloride after rapid freezing and freeze-substitution fixation.
T Hatae et al.
Journal of electron microscopy, 33(2), 186-190 (1984-01-01)
Guoguang Liu et al.
Biomacromolecules, 9(3), 949-953 (2008-02-15)
The feasibility of a previously established method based on ozonolysis and hydrogenation reactions for the production of 9-hydroxynonanoic acid from oleic acid has been demonstrated. Metal catalyzed lactonization conditions have been used to convert 9-hydroxynonanoic acid into 1,11-dioxacycloicosane-2,12-dione, which is
Lutz Ackermann et al.
Organic & biomolecular chemistry, 5(12), 1975-1978 (2007-06-07)
Two distinct economical catalysts for intramolecular hydroaminations of electronically unactivated alkenes with basic amines are described, which are based on (a) group 4 metal halides under basic reaction conditions or (b) Brønsted-acid organocatalysts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service