Skip to Content
Merck
All Photos(1)

Key Documents

8.22337

Sigma-Aldrich

m-Xylene

for synthesis

Synonym(s):

m-Xylene, 1,3-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-576-3
NACRES:
NA.22

vapor pressure

8 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

525 °C

expl. lim.

1.1-7 % (v/v)

bp

139.1 °C/1013 hPa

mp

-48 °C

transition temp

flash point 27 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3

InChI key

IVSZLXZYQVIEFR-UHFFFAOYSA-N

General description

m-Xylene is an aromatic hydrocarbon primarily used as a solvent in various industrial applications, such as in the production of coatings, paints, rubber products, and adhesives. Additionally, m-xylene serves as a crucial precursor in the production of certain chemicals like phthalic anhydride and isophthalic acid.

Application

m-Xylene can be used as a solvent in the:
  • Formation of vertical nanowires composed of C60 molecules through the solvent vapor annealing (SVA) process.
  • Copper-catalyzed halogen exchange reaction in aryl halides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Percutaneous absorption of m-xylene in man
Engstrom K, et al.},
International Archives of Occupational and Environmental Health, 39, 181-189 (1977)
A Study on the Production of Isophthalic Acid from M-xylene under the Catalysis of Cobalt and H3PW12O40/Carbon Modified by HNO3 Solution
Wang ZH, et al.},
International Journal of Chemical Reactor Engineering, 13(3), 413-425 (2015)
Jungah Kim et al.
ACS nano, 7(10), 9122-9128 (2013-09-11)
We report that C60 molecules are spontaneously crystallized into vertical nanowires by the solvent vapor annealing (SVA) process. C60 molecules have been known to be assembled into wire-like crystals by simply dropping and drying C60 solutions in m-xylene on a
Characterization of products formed from the oxidation of toluene and m-xylene with varying NOx and OH exposure
Srivastava D, et al.
Chemosphere, 334, 139002-139002 (2023)
Artis Klapars et al.
Journal of the American Chemical Society, 124(50), 14844-14845 (2002-12-12)
A mild and general method for the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides was developed utilizing a catalyst system comprising 5 mol % of CuI and 10 mol % of a 1,2- or 1,3-diamine ligand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service