Skip to Content
Merck
All Photos(2)

Key Documents

850375C

Avanti

18:1 (Δ9-Cis) PC (DOPC)

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; DOPC; PC(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84NO8P
CAS Number:
Molecular Weight:
786.11
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dioleoyl-sn-glycero-3-phosphocholine, chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850375C-25mg)
pkg of 2 × 20 mL (850375C-1g)
pkg of 2 × 4 mL (850375C-200mg)
pkg of 5 × 4 mL (850375C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (850375C-25mg)
25 mg/mL (850375C-1g)
25 mg/mL (850375C-200mg)
25 mg/mL (850375C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCC\C=C/CCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C44H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,42H,6-19,24-41H2,1-5H3/b22-20-,23-21-/t42-/m0/s1

InChI key

SNKAWJBJQDLSFF-AOMFJDGTSA-N

General description

The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

18:1 (Δ9-Cis) PC (DOPC) has been used in the preparation of planar supported lipid bilayer for fluorescence microscopy. It is also suitable for use in the preparation of protocells.

Biochem/physiol Actions

18:1 (Δ9-Cis) PC (DOPC) shows high resistance to the toxic effects of dimethyl sulfoxide (DMSO).

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850375C-1g)
5 mL Clear Glass Sealed Ampule (850375C-200mg)
5 mL Clear Glass Sealed Ampule (850375C-25mg)
5 mL Clear Glass Sealed Ampule (850375C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular dynamics simulations of the interactions of DMSO with DPPC and DOPC phospholipid membranes
Hughes ZE, et al.
The Journal of Physical Chemistry B, 116(39), 11911-11923 (2012)
K-Ras4B remains monomeric on membranes over a wide range of surface densities and lipid compositions
Chung JK, et al.
Biophysical Journal, 114(1), 137-145 (2018)
Mesoporous silica-supported lipid bilayers (protocells) for DNA cargo delivery to the spinal cord
Dengler EC, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 168(2), 209-224 (2013)
Yachong Guo et al.
ACS nano (2018-11-20)
Increasing awareness of bioeffects and toxicity of nanomaterials interacting with cells puts in focus the mechanisms by which nanomaterials can cross lipid membranes. Apart from well-discussed energy-dependent endocytosis for large objects and passive diffusion through membranes by solute molecules, other
Tatiana K Rostovtseva et al.
The Journal of biological chemistry, 287(35), 29589-29598 (2012-07-06)
Elucidating molecular mechanisms by which lipids regulate protein function within biological membranes is critical for understanding the many cellular processes. Recently, we have found that dimeric αβ-tubulin, a subunit of microtubules, regulates mitochondrial respiration by blocking the voltage-dependent anion channel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service