Skip to Content
Merck
All Photos(2)

Key Documents

850355C

Avanti

16:0 PC (DPPC)

Avanti Research - A Croda Brand

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphocholine; DPPC; PC(16:0/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dipalmitoyl-sn-glycero-3-phosphocholine, chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850355C-25mg)
pkg of 1 × 20 mL (850355C-500mg)
pkg of 2 × 20 mL (850355C-1g)
pkg of 2 × 4 mL (850355C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (850355C-25mg)
25 mg/mL (850355C-1g)
25 mg/mL (850355C-200mg)
25 mg/mL (850355C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C40H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-39(42)46-36-38(37-48-50(44,45)47-35-34-41(3,4)5)49-40(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3

InChI key

KILNVBDSWZSGLL-UHFFFAOYSA-N

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes. It is also an important component of the mucosal layer of the colon.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

16:0 PC (DPPC) is suitable for use:
  • in vesicle preparation
  • as the main phospholipid component for all ultrasound contrast agents (UCA) formulations
  • as a component for microbubble preparation
  • in the preparation of lipid bilayers

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance. It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850355C-1g)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850355C-500mg)
5 mL Clear Glass Sealed Ampule (850355C-200mg)
5 mL Clear Glass Sealed Ampule (850355C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integration and oligomerization of Bax protein in lipid bilayers characterized by single molecule fluorescence study
Luo L, et al.
The Journal of Biological Chemistry, 289(46), 31708-31718 (2014)
Protein aggregation with poly (vinyl) alcohol surfactant reduces double emulsion-encapsulated mammalian cell-free expression
Ho KKY, et al.
PLoS ONE, 12(3), e0174689-e0174689 (2017)
Constructing droplet interface bilayers from the contact of aqueous droplets in oil
Leptihn S, et al.
Nature Protocols, 8(6), 1048-1048 (2013)
Microbubble composition and preparation for high-frequency contrast-enhanced ultrasound imaging: in vitro and in vivo evaluation
Daeichin V, et al.
IEEE Transactions on Ultrasonics, Ferroelectrics, and Frequency Control, 64(3), 555-567 (2016)
S Q Choi et al.
Nature communications, 2, 312-312 (2011-05-19)
Two-dimensional films of surface-active agents-from phospholipids and proteins to nanoparticles and colloids-stabilize fluid interfaces, which are essential to the science, technology and engineering of everyday life. The 2D nature of interfaces present unique challenges and opportunities: coupling between the 2D

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service