Skip to Content
Merck
All Photos(1)

Key Documents

860516P

Avanti

C16 Ceramide (d18:1/16:0)

Avanti Research - A Croda Brand

Synonym(s):

N-palmitoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H67NO3
CAS Number:
Molecular Weight:
537.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860516P-10mg)
pkg of 1 × 100 mg (860516P-100mg)
pkg of 1 × 25 mg (860516P-25mg)
pkg of 1 × 5 mg (860516P-5mg)
pkg of 1 × 50 mg (860516P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C34H67NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33(37)32(31-36)35-34(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h27,29,32-33,36-37H,3-26,28,30-31H2,1-2H3,(H,35,38)/b29-27+/t32-,33+/m0/s1

InChI key

YDNKGFDKKRUKPY-TURZORIXSA-N

General description

Ceramide comprise sphingosine and fatty acid and belongs to the sphingolipids family.

Application

C16 Ceramide (d18:1/16:0) has been used:
  • as an internal standard in the chromatography with negative ion electrospray ionization coupled to tandem mass spectrometry for myocardial ceramide quantification
  • as an internal standard for the ceramide quantification from rat samples using liquid chromatography-mass spectrometry
  • to test its effect on mitochondrial stress response in worms

Biochem/physiol Actions

C16 Ceramide levels are elevated in chronic alcohol-related liver disease (ALD). In general, ceramide inhibits mitochondrial function and insulin signaling. They are essential component of cell membrane and are involved in lipid raft formation. C16 ceramide based analog (C2-C16 CCPS) is potential anticancer agent used in chemotherapy. The levels of C16 ceramide and its ratio with other ceramides is regarded as a biomarker in predicting coronary artery disease (CAD). It interacts with the DNA-binding domain of tumor suppressor protein p53 and activates p53.

Packaging

5 mL Amber Glass Screw Cap Vial (860516P-100mg)
5 mL Amber Glass Screw Cap Vial (860516P-10mg)
5 mL Amber Glass Screw Cap Vial (860516P-25mg)
5 mL Amber Glass Screw Cap Vial (860516P-50mg)
5 mL Amber Glass Screw Cap Vial (860516P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aritz B García-Arribas et al.
Langmuir : the ACS journal of surfaces and colloids, 32(35), 9053-9063 (2016-08-04)
The effects of increasing amounts of palmitoylceramide (pCer) on human red blood cell lipid membranes have been studied using atomic force microscopy of supported lipid bilayers, in both imaging (bilayer thickness) and force-spectroscopy (nanomechanical resistance) modes. Membranes appeared homogeneous with
M Laura Fanani et al.
Langmuir : the ACS journal of surfaces and colloids, 34(39), 11749-11758 (2018-09-06)
Sphingosine [(2 S,3 R,4 E)-2-amino-4-octadecene-1,3-diol] is the most common sphingoid base in mammals. Ceramides are N-acyl sphingosines. Numerous small variations on this canonical structure are known, including the 1-deoxy, the 4,5-dihydro, and many others. However, whenever there is a Δ4
Insulin resistance, ceramide accumulation, and endoplasmic reticulum stress in human chronic alcohol-related liver disease
Longato L, et al.
Oxidative Medicine and Cellular Longevity, 2012 (2012)
Development and validation of a high-throughput LC-MS/MS assay for routine measurement of molecular ceramides
Kauhanen D, et al.
Analytical and Bioanalytical Chemistry, 408(13), 3475-3483 (2016)
Meenu N Perera et al.
Chemistry and physics of lipids, 194, 110-116 (2015-09-27)
A ceramide commonly found in mammalian cells, C16-ceramide (N-palmitoyl-d-erythro-sphingosine), is capable of forming large, protein-permeable channels in the mitochondrial outer membrane (MOM). However, C16-ceramide is unable to permeabilize the plasma membrane of erythrocytes. This specificity is unexpected considering that ceramide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service