Skip to Content
Merck
All Photos(2)

Key Documents

840875C

Avanti

18:1 PA

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphate (sodium salt); DOPA; PA(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O8PNa
CAS Number:
Molecular Weight:
722.95
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dioleoyl-sn-glycero-3-phosphate (sodium salt), chloroform solution

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840875C-25mg)
pkg of 2 × 20 mL (840875C-1g)
pkg of 2 × 4 mL (840875C-200mg)
pkg of 5 × 4 mL (840875C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (840875C-25mg)
25 mg/mL (840875C-1g)
25 mg/mL (840875C-200mg)
25 mg/mL (840875C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na].[P](=O)(OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCC\C=C/CCCCCCCC)(O)O

InChI

1S/C39H73O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,37H,3-16,21-36H2,1-2H3,(H2,42,43,44);/b19-17-,20-18-;/t37-;/m1./s1

InChI key

KHDHADSEKGCPKW-ZBFGHDQJSA-N

General description

18:1 PA (1,2-dioleoyl-sn-glycero-3-phosphate (sodium salt)) is a sodium salt of modified lipid which has phosphatidic acid group as the backbone.

Application

18:1 PA (1,2-dioleoyl-sn-glycero-3-phosphate (sodium salt)) has been used:
  • as a synthetic standard to identify the position of PA after iodine staining in phosphatidic acid assay by thin-layer chromatography (TLC)
  • as a lipid standard in the optimization of the liquid chromatography−mass spectrometry (LC-MS) method conditions
  • as a lipid standard in radiolabeling analysis of phospholipids

Biochem/physiol Actions

Phosphatidic acid (PA) serves as a lipid messenger. It plays a key role in mammalian target of rapamycin (mTOR) signaling.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (840875C-1g)
5 mL Clear Glass Sealed Ampule (840875C-200mg)
5 mL Clear Glass Sealed Ampule (840875C-25mg)
5 mL Clear Glass Sealed Ampule (840875C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinhui Sun et al.
Cell cycle (Georgetown, Tex.), 14(5), 721-731 (2015-01-16)
It had been known for decades that primordial follicles in mammalian ovaries are assembled with definite numbers and represent the ovarian reserve throughout the reproductive life. Intra-oocyte PI3K/mTOR pathways have been indicated to play a central role on the activation
Shutao Guo et al.
Journal of controlled release : official journal of the Controlled Release Society, 174, 137-142 (2013-11-28)
The poor solubility of cisplatin (CDDP) often presents a major obstacle in the formulation of CDDP in nanoparticles (NPs) by traditional methods. We have developed a novel method for synthesizing CDDP NPs taking advantage of its poor solubility. By mixing
Kathleen Frondorf et al.
The Journal of biological chemistry, 285(21), 15837-15847 (2010-03-23)
Phosphatidic acid (PA) is a pleiotropic lipid second messenger in mammalian cells. We report here that extracellular PA acts as a leukocyte chemoattractant, as membrane-soluble dioleoyl-PA (DOPA) elicits actin polymerization and chemotaxis of human neutrophils and differentiated proleukemic HL-60 cells.
Reika Tei et al.
The Journal of cell biology, 219(3) (2020-01-31)
Phosphatidic acid (PA) is both a central phospholipid biosynthetic intermediate and a multifunctional lipid second messenger produced at several discrete subcellular locations. Organelle-specific PA pools are believed to play distinct physiological roles, but tools with high spatiotemporal control are lacking
Zhenhua Hu et al.
Science advances, 6(14), eaba0145-eaba0145 (2020-04-10)
Low oral bioavailability of peptide drugs has limited their application to parenteral administration, which suffers from poor patient compliance. Here, we show that molecular targeting of the FATP4 transporter is an effective approach to specifically transport long-chain fatty acid (LCFA)-conjugated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service