Skip to Content
Merck
All Photos(2)

Key Documents

850356C

Avanti

4ME 16:0 PC

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(3,7,11,15-tetramethylhexadecanoyl)-sn-glycero-3-phosphocholine; PC(16:0(3me,7me,11me,15me)/16:0(3me,7me,11me,15me))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H96NO8P
CAS Number:
Molecular Weight:
846.25
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-diphytanoyl-sn-glycero-3-phosphocholine, chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850356C-25mg)
pkg of 1 × 20 mL (850356C-500mg)
pkg of 2 × 4 mL (850356C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (850356C-25mg)
25 mg/mL (850356C-200mg)
25 mg/mL (850356C-500mg)

shipped in

dry ice

storage temp.

−20°C

General description

4ME 16:0 PC/DPhPC (1,2-diphytanoyl-sn-glycero-3-phosphocholine is a synthetic lipid. It has ester linkages between acyl chains and glycerol backbone and also between the headgroup and glycerol backbone. This neutral lipids possess high steric asymmetry. It remains in fluid or viscous liquid crystalline state over a wide range of temperature.
Lipids containing diphytanoyl fatty acid chains have been used to produce stable planar lipid membranes (see References). Diphytanoyl phosphatidylcholine does not exhibit a detectable gel to liquid crystalline phase transition from -120°C to +120°C.The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products from oxidization and hydrolysis.

Application

4ME 16:0 PC is suitable for use:
  • to study its phase behaviour and its interactions with the representative antimicrobial peptide PGLa from Xenopus laevis (peptidyl-glycylleucine-carboxyamide
  • to improve the mechanical stability of planar lipid bilayer membrane
  • to prepare giant unilamellar vesicles (GUVs)

Biochem/physiol Actions

4ME 16:0 PC/DPhPC (1,2-diphytanoyl-sn-glycero-3-phosphocholine can be used as a bilayer mimic for electrophysiological measurements. It is stable under mechanical stress but the ester linkages are liable to hydrolysis under acidic or alkaline conditions.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850356C-500mg)
5 mL Clear Glass Sealed Ampule (850356C-200mg)
5 mL Clear Glass Sealed Ampule (850356C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation and characterization of planar lipid bilayer membranes from synthetic phytanyl-chained glycolipids
Baba T, et al.
Biochimica et Biophysica Acta - Biomembranes, 1421(1), 91-102 (1999)
H Lindsey et al.
Biochimica et biophysica acta, 555(1), 147-167 (1979-07-19)
We report here on a series of studies aimed at characterization of the structural and dynamical properties of the synthetic lipid diphytanoyl phosphatidylcholine, in multilamellar dispersions and vesicle suspensions. The lipid exhibits no detectable gel to liquid crystalline phase transition
Patrick Urban et al.
Langmuir : the ACS journal of surfaces and colloids, 34(44), 13368-13374 (2018-10-23)
Controlling lateral interactions between lipid molecules in a bilayer membrane to guide membrane organization and domain formation is a key factor for studying and emulating membrane functionality in synthetic biological systems. Here, we demonstrate an approach to reversibly control lipid
Zahra Aminipour et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 146, 133-142 (2019-11-08)
Doxorubicin is an anti-cancer drug that is important for breast cancer therapy. In this study, the effects of the membrane potential of breast cancer cells (-30 mV) and normal breast epithelial cells (-60 mV) on doxorubicin (DOX) permeability was studied. To achieve
Tim Diederichs et al.
Nature communications, 10(1), 5018-5018 (2019-11-07)
Nanopores are key in portable sequencing and research given their ability to transport elongated DNA or small bioactive molecules through narrow transmembrane channels. Transport of folded proteins could lead to similar scientific and technological benefits. Yet this has not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service