Skip to Content
Merck
All Photos(2)

Key Documents

840857C

Avanti

16:0-18:1 PA

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1-hexadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphate (sodium salt); POPA; PA(16:0/18:1(9Z)); 110616

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H70O8PNa
CAS Number:
Molecular Weight:
696.91
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840857C-25mg)
pkg of 2 × 4 mL (840857C-200mg)
pkg of 5 × 4 mL (840857C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840857C

concentration

10 mg/mL (840857C-25mg)
25 mg/mL (840857C-200mg)
25 mg/mL (840857C-500mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H71O8P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(39)45-35(34-44-46(40,41)42)33-43-36(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,35H,3-16,19-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/b18-17-;/t35-;/m1./s1

InChI key

PPYXMAJBOKWTQX-RYRMQHSSSA-M

General description

16:0-18:1 PA (1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt)/POPA) has eight oxygen atoms and one hydroxyl hydrogen atom. POPA is an anionic lipid.

Application

16:0-18:1 PA (1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt)) has been used to study its effects on the monogalactosyldiacylglycerol (MGDG) synthase activity of chloroplast envelope and in liposome Pull-Down Assay with the C2-like domain of SEIPIN.

Biochem/physiol Actions

Phosphatidic acid (PA) serves as a precursor for phosphatidylcholine or phosphatidylserine. It is involved in influencing the membrane curvature and signaling.

Packaging

5 mL Clear Glass Sealed Ampule (840857C-200mg)
5 mL Clear Glass Sealed Ampule (840857C-25mg)
5 mL Clear Glass Sealed Ampule (840857C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allison Dickey et al.
Biophysical journal, 95(6), 2636-2646 (2008-06-03)
To better understand bilayer property dependency on lipid electrostatics and headgroup size, we use atomistic molecular dynamics simulations to study negatively charged and neutral lipid membranes. We compare the negatively charged phosphatidic acid (PA), which at physiological pH and salt
Renhong Yan et al.
Developmental cell, 47(2), 248-256 (2018-10-09)
The biogenesis of lipid droplets (LDs) and the development of adipocytes are two key aspects of mammalian fat storage. SEIPIN, an integral membrane protein of the endoplasmic reticulum (ER), plays a critical role in both LD formation and adipogenesis. The
Nadezhda Y Davydova et al.
The Biochemical journal, 476(23), 3661-3685 (2019-11-22)
In this study, we investigate the ability of ethanol-inducible CYP2E1 to interact with other cytochrome P450 species and affect the metabolism of their substrates. As a model system, we used CYP2E1-enriched human liver microsomes (HLM) obtained by the incorporation of
R J Perrin et al.
The Journal of biological chemistry, 275(44), 34393-34398 (2000-08-23)
alpha-Synuclein has been centrally implicated in neurodegenerative disease, and a normal function in developmental synaptic plasticity has been suggested by studies in songbirds. A variety of observations suggest the protein partitions between membrane and cytosol, a behavior apparently conferred by
Tingting Wang et al.
Frontiers in endocrinology, 9, 221-221 (2018-05-22)
Searching for nutraceuticals and understanding the underlying mechanism that promote fish growth is at high demand for aquaculture industry. In this study, the modulatory effects of soy phosphatidic acids (PA) on cell proliferation, nutrient sensing, and metabolic pathways were systematically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service