Skip to Content
Merck
All Photos(4)

Key Documents

610000

Avanti

Extruder Set with Holder/Heating Block

Avanti Research - A Croda Brand

Synonym(s):

Mini-Extruder Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151503
NACRES:
NB.51

feature

Extruder Set with Block

manufacturer/tradename

Avanti Research - A Croda Brand

General description

The Avanti Mini-Extruder allows researchers and scientists to prepare large, unilamellar vesicles by extrusion (LUVET) in an efficient, rapid manner.

Explore the instructions to learn more about the Avanti Research Mini-Extruder.

Application

Extruder Set with Holder/Heating Block has been used for the preparation of vesicles and liposomes.

Packaging

Mini-Extruder (set): includes instructions, mini-extruder, two 1000μL syringes, 100 pore size (0.1μm) polycarbonate membranes, 100 filter supports, and 1 holder/heating block

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 610020Mini-Extruder (only), Avanti Research - A Croda BrandSDS

  • 610017Syringe (1000 μL), Avanti Research - A Croda BrandSDS

  • 610014Filter Support, Avanti Research - A Croda BrandSDS

  • 610005PC Membranes 0.1um, Avanti Research - A Croda BrandSDS

  • 610024Extr. Heating Block, Avanti Research - A Croda BrandSDS

also commonly purchased with this product

Product No.
Description
Pricing

recommended

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liposome-enhanced lateral-flow assays for clinical analyses
Edwards KA, et al.
Biosensors and Biodetection, 407-434 (2017)
Immobilization and detection of platelet-derived extracellular vesicles on functionalized silicon substrate: cytometric and spectrometric approach
Gajos K, et al.
Analytical and Bioanalytical Chemistry, 409(4), 1109-1119 (2017)
Trapping single molecules in liposomes: surface interactions and freeze- thaw effects
Liu B, et al.
The Journal of Physical Chemistry B, 114(46), 15191-15198 (2010)
In vivo experimental investigation on multi-pulse Nd: YAG laser-induced vesicle embolization of vascular lesions
Jia H, et al.
Optics & Laser technology, 116, 246-255 (2019)

Protocols

Watch the below video for instructions on liposome preparation using the Avanti® Mini-Extruder.

Watch the below video for instructions on liposome preparation using the Avanti® Mini-Extruder.

Watch the below video for instructions on liposome preparation using the Avanti® Mini-Extruder.

Watch the below video for instructions on liposome preparation using the Avanti® Mini-Extruder.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service