Skip to Content
Merck
All Photos(2)

Key Documents

840457P

Avanti

16:0-18:1 PG

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); POPG; PG(16:0/18:1(9Z)); 110650

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H76O10PNa
CAS Number:
Molecular Weight:
770.99
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840457P-1g)
pkg of 2 × 100 mg (840457P-200mg)
pkg of 5 × 100 mg (840457P-500mg)
pkg of 1 × 25 mg (840457P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H77O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,37-38,41-42H,3-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b18-17-;/t37?,38-;/m1./s1

InChI key

FJXDNGDRHUDFST-XQYKCTAGSA-M

General description

Phosphoglyceride (PG) are also referred as glycerophospholipid. It is an important component of membrane bilayers. It has three-carbon backbone of glycerol, two long-chain fatty acids, esterified to hydroxyl groups on carbons 1 and 2 of the glycerol and phosphoric acid esterified to the C3 hydroxyl group of glycerol.[1] It is found at high levels in the membranes of all cells and at low levels in fat stores.[2]

Application

16:0-18:1 PG has been used in the preparation of large unilamellar vesicles (LUVs).[3]

Biochem/physiol Actions

Phosphoglycerides (PG)/glycerophospholipids play a key role in cell signaling systems. It also serves as an anchor for proteins in cell membranes.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840457P-200mg)
5 mL Clear Glass Sealed Ampule (840457P-25mg)
5 mL Clear Glass Sealed Ampule (840457P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (840457P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane Structure and Dynamics
Cell Biology, 227- 239 (2017)
Lipids
Medical Biochemistry, 283(18), 99- 119 (2017)
Sandeep Kumar et al.
The journal of physical chemistry. B, 122(26), 6763-6770 (2018-06-08)
The interaction of amphiphilic ionic liquids containing an 1-alkyl-3-methylimidazolium cation ([C12MIM]+), which shows acute cytotoxicity toward marine and bacterial life, with zwitterionic 1-palmitoyl-2-oleoyl- sn-glycero-3-phospho-choline (POPC) and anionic 1-palmitoyl-2-oleoyl- sn-glycero-3-phospho- rac-glycerol (POPG) membranes was investigated. Biophysical parameters of this interaction were
Takeshi Sunami et al.
Langmuir : the ACS journal of surfaces and colloids, 34(1), 88-96 (2017-12-08)
Liposome fusion is a way of supplying additional components for in-liposome biochemical reactions. Electrofusion is a method that does not require the addition of fusogens, which often alter the liposome dispersion, and is therefore useful for repetitive liposome fusion. However
Tânia Silva et al.
Langmuir : the ACS journal of surfaces and colloids, 34(5), 2158-2170 (2018-01-07)
An understanding of the mechanism of action of antimicrobial peptides is fundamental to the development of new and more active antibiotics. In the present work, we use a wide range of techniques (SANS, SAXD, DSC, ITC, CD, and confocal and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service