Skip to Content
Merck
All Photos(2)

Key Documents

730408

Sigma-Aldrich

1-Allyl-2-bromobenzene

95%

Synonym(s):

2-Allylbromobenzene, 2-Allylphenyl bromide, 2-Bromo-1-allylbenzene, o-Allylbromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9Br
CAS Number:
Molecular Weight:
197.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.559

density

1.311 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Brc1ccccc1CC=C

InChI

1S/C9H9Br/c1-2-5-8-6-3-4-7-9(8)10/h2-4,6-7H,1,5H2

InChI key

NTYPQDZCDDLKLV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.9 °F

Flash Point(C)

93.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service