Skip to Content
Merck
All Photos(1)

Key Documents

383406

Sigma-Aldrich

5′-Bromo-2′-hydroxyacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(OH)COCH3
CAS Number:
Molecular Weight:
215.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

58-61 °C (lit.)

functional group

bromo
ketone

SMILES string

CC(=O)c1cc(Br)ccc1O

InChI

1S/C8H7BrO2/c1-5(10)7-4-6(9)2-3-8(7)11/h2-4,11H,1H3

InChI key

HQCCNFFIOWYINW-UHFFFAOYSA-N

Application

5′-Bromo-2′-hydroxyacetophenone (5-Bromo-2-hydroxyacetophenone) may be used in the following studies:
  • As ligand in the preparation of tetrahedral metallocene complexes containing vanadium(IV) (vanadocene), having potential spermicidal activity against human sperm.
  • Synthesis of {2′-[1-(5-bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris(pyridine) nickel(II)] pyridine solvate.
  • Synthesis of N′-[1-(5-bromo-2-hydroxyphenyl)ethylidene]-3,4,5-trihydroxybenzohydrazide dimethyl sulfoxide solvate trihydrate.
  • Preparation of 6-bromochromen-4-one.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5'-Bromo-2'-hydroxyacetophenone.
Ng SW.
Acta Crystallographica Section E, Structure Reports Online, 63(4), o1805-o1806 (2007)
{2'-[1-(5-Bromo-2-oxidophenyl) ethylidene] benzohydrazidato (2-)} tris (pyridine) nickel (II)] pyridine solvate.
Ali HM, et al.
Acta Crystallographica Section E, Structure Reports Online, 60(11), m1708-m1709 (2004)
O J D'Cruz et al.
Biology of reproduction, 60(2), 435-444 (1999-01-23)
We have recently reported that tetrahedral metallocene complexes containing vanadium(IV) (vanadocene) have potent spermicidal activity against human sperm. The spermicidal activity was dependent on vanadium(IV) as the central metal ion within the bis-cyclopentadienyl (Cp2)-metal complex, but the variation of diacido
Nura Suleiman Gwaram et al.
Acta crystallographica. Section E, Structure reports online, 66(Pt 3), o721-o721 (2010-01-01)
The benzohydrazide mol-ecule in the title compound, C(15)H(13)BrN(2)O(5)·C(2)H(6)OS·3H(2)O, is almost planar with an r.m.s. deviation for the non-H atoms of 0.078 Å. The organic mol-ecules, water and dimethyl sulfoxide solvent mol-ecules are linked by N-H⋯O, O-H⋯O and O-H⋯S inter-molecular hydrogen bonds
Justin J J Leahy et al.
Bioorganic & medicinal chemistry letters, 14(24), 6083-6087 (2004-11-18)
A solution-phase multiple-parallel synthesis approach was employed for the preparation of 6-, 7- and 8-aryl-substituted chromenone libraries, which were screened as inhibitors of the DNA repair enzyme DNA-dependent protein kinase (DNA-PK). These studies resulted in the identification of 8-dibenzothiophen-4-yl-2-morpholin-4-yl-chromen-4-one (NU7441)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service