Skip to Content
Merck
All Photos(3)

Key Documents

417599

Sigma-Aldrich

4-Methoxyphenylboronic acid

≥95.0%

Synonym(s):

(4-Methoxyphenyl)boric acid, (p-Methoxyphenyl)boronic acid, 4-Anisylboronic acid, 4-Methoxybenzeneboronic acid, p-Anisylboronic acid, p-Methoxybenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4B(OH)2
CAS Number:
Molecular Weight:
151.96
Beilstein:
2936912
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

form

powder

mp

204-206 °C (lit.)

SMILES string

COc1ccc(cc1)B(O)O

InChI

1S/C7H9BO3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5,9-10H,1H3

InChI key

VOAAEKKFGLPLLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used for
  • Suzuki-Miyaura cross-coupling reactions
  • Pd-catalyzed direct arylation
  • Highly effective synthesis using palladium-catalyzed arylation Suzuki-Miyaura cross-coupling in water
  • Palladium-catalyzed stereoselective Heck-type reaction
  • Tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence
  • Copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides
  • Ruthenium catalyzed direct arylation
  • Rh-catalyzed asymmetric conjugate addition
  • Ligand-free copper-catalyzed coupling

Reagent used in Preparation of
  • Palladium(II) thiocarboxamide complexes as Suzuki coupling catalyst
  • Push-pull arylvinyldiazine chromophores with photophysical properties

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosuke Ishijima et al.
Dalton transactions (Cambridge, England : 2003), 49(44), 15612-15621 (2020-09-24)
Benzo[b]arsole derivatives, being arsenic analogues of indole, were synthesized by utilizing a safely prepared arsenic precursor. The structural and photophysical properties of the obtained 2-arylbenzo[b]arsoles were experimentally and computationally studied in comparison with those of 1,2,5-triarylarsoles and 9-phenylarsafluorene. It was
Mikhail Y Vorona et al.
Materials (Basel, Switzerland), 12(17) (2019-08-29)
Anthracene-based semiconductors are a class of molecules that have attracted interest due to their air stability, planarity, potential for strong intermolecular interactions, and favorable frontier molecular orbital energy levels. In this study seven novel 9,10-anthracene-based molecules were synthesized and their
M Lakshmi Kantam et al.
The Journal of organic chemistry, 71(25), 9522-9524 (2006-12-02)
N-Arylation of imidazoles and amines with arylboronic acids was accomplished with copper-exchanged fluorapatite (CuFAP) in methanol at room temperature. The products N-arylimidazoles and N-arylamines were isolated in good to excellent yields. A variety of arylboronic acids were converted to the
Maria Makrinich et al.
Solid state nuclear magnetic resonance, 84, 196-203 (2017-05-06)
The ability of various pulse types, which are commonly applied for distance measurements, to saturate or invert quadrupolar spin polarization has been compared by observing their effect on magnetization recovery curves under magic-angle spinning. A selective central transition inversion pulse
Sylvain Achelle et al.
The Journal of organic chemistry, 77(8), 4087-4096 (2012-04-06)
A new series of push-pull arylvinyldiazines has been efficiently prepared by aldol condensation between the appropriate methyldiazine and aromatic aldehyde. The optical absorption and emission properties of these chromophores were studied in different solvents and media. These compounds act as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service