Skip to Content
Merck
All Photos(1)

Key Documents

A29402

Sigma-Aldrich

Allylbenzene

98%

Synonym(s):

3-Phenyl-1-propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH=CH2
CAS Number:
Molecular Weight:
118.18
Beilstein:
1098501
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

156-157 °C (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

C=CCc1ccccc1

InChI

1S/C9H10/c1-2-6-9-7-4-3-5-8-9/h2-5,7-8H,1,6H2

InChI key

HJWLCRVIBGQPNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Allylbenzene, also known as 3-phenylpropene, is an organic compound used as a source of the allyl group in allylation reactions.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eric A Standley et al.
Journal of the American Chemical Society, 135(4), 1585-1592 (2013-01-16)
The synthesis and characterization of the air-stable nickel(II) complex trans-(PCy(2)Ph)(2)Ni(o-tolyl)Cl is described in conjunction with an investigation of its use for the Mizoroki-Heck-type, room temperature, internally selective coupling of substituted benzyl chlorides with terminal alkenes. This reaction, which employs a
Mitsuo Miyazawa et al.
Natural product research, 19(1), 29-36 (2005-02-11)
A methanol extract from Asiasarum heterotropoides showed a suppressive effect of the SOS-including activity on the mutagen 2-amino-3,4-dimethylimidazo[4,5-f]quinoline (MeIQ) in the Salmonella typhimurium TA1535/pSK1002 umu test. The methanol extract was re-extracted with chloroform, butanol, and water. The chloroform fraction showed
J R Idle
Prague medical report, 106(1), 27-38 (2005-07-13)
The typical spices used in winter include nutmeg, cinnamon, clove and anise. These spices contain two groups of chemicals, the allylbenzenes and their isomers, the propenylbenzenes. It was suggested 40 years ago by Alexander Shulgin that these substances act as
Thi X Thi Luu et al.
Molecules (Basel, Switzerland), 14(9), 3411-3424 (2009-09-29)
Essential oil allylbenzenes from have been converted quickly and efficiently into the corresponding benzaldehydes in good yields by a two-step "green" reaction pathway based on a solventless alkene group isomerization by KF/Al(2)O(3) to form the corresponding 1-arylpropene and a subsequent
P G Debrunner et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(23), 12791-12798 (1996-11-12)
We have used Mössbauer and electron paramagnetic resonance (EPR) spectroscopy to study a heme-N-alkylated derivative of chloroperoxidase (CPO) prepared by mechanism-based inactivation with allylbenzene and hydrogen peroxide. The freshly prepared inactivated enzyme ("green CPO") displayed a nearly pure low-spin ferric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service