Skip to Content
Merck
All Photos(3)

Key Documents

205869

Sigma-Aldrich

Palladium(II) acetate

reagent grade, 98%

Synonym(s):

Pd(OAc)2, [Pd(OAc)2]3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(OCOCH3)2
CAS Number:
Molecular Weight:
224.51
Beilstein:
6086766
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

Assay

98%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

216.3-223.7 °C (dec.)

SMILES string

CC(O[Pd]OC(C)=O)=O

InChI

1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

YJVFFLUZDVXJQI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

[Pd(OAc)2]3 is a palladium coordination complex and a heterogenous metal catalyst that is widely used in organic synthesis such as transmetalation, insertion, oxidative addition, direct homocoupling of aryl halides, Buchwald-Hartwig reaction of C-N bond formation, reduction of alkynes, and reductive elimination reactions. It is also employed as a starting material for the synthesis of other Pd(II) compounds as well as for the preparation of active palladium catalysts for Suzuki-Miyaura cross-coupling and C-H functionalization reactions.

Application

[Pd(OAc)2]3 can be used as a:
  • Catalyst for the regioselective anti-hydrochlorination of the terminal and internal alkynes.
  • Precursor to prepare a heterogeneous palladium complex catalyst for the Heck-Coupling reaction and Sonogashira cross-coupling reaction.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of bis (amine) palladium (ii) carboxylate complexes as precursors of palladium nanoparticles
Zakrzewska J and Uznanski P
Dalton Transactions, 50, 6933-6948 (2021)
Polystyrene-Anchored Palladium (II) Schiff Base Complex: A Reusable Catalyst for Phosphine-Free and Copper-Free Sonogashira Cross-Coupling Reaction in Aqueous Medium
Islam M, et al.
Synthetic Communications, 41, 2583-2593 (2011)
Mesoporous Palladium N, N?-Bis (3-Allylsalicylidene) o-Phenylenediamine-Methyl Acrylate Resins as Heterogeneous Catalysts for the Heck Coupling Reaction
Mella C, et al.
MATERIALS, 12, 2612-2612 (2019)
Palladium (II) Acetate
Grennberg H, et al
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Palladium (II) Acetate [Pd (OAc)2]: A Versatile Catalyst
Vats R
Synlett, 2006, 329-330 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service