Skip to Content
Merck
All Photos(1)

Key Documents

58035

Sigma-Aldrich

(3-Iodopropyl)trimethoxysilane

≥95.0% (GC)

Synonym(s):

3-Iodo-n-propyl-1-trimethoxysilane, Iodopropyltrimethoxysilane, Trimethoxy(iodopropyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15IO3Si
CAS Number:
Molecular Weight:
290.17
Beilstein:
969628
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

refractive index

n20/D 1.473 (lit.)

density

1.482 g/mL at 20 °C (lit.)

functional group

iodo

SMILES string

CO[Si](CCCI)(OC)OC

InChI

1S/C6H15IO3Si/c1-8-11(9-2,10-3)6-4-5-7/h4-6H2,1-3H3

InChI key

NILZGRNPRBIQOG-UHFFFAOYSA-N

Application

(3-iodopropyl)trimethoxysilane may be used to organically modify mesoporous silica support by grafting method.{85} The product may be used to synthesize heavy atom-concentrated organically modified silica nanoparticles.{86}

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goutam Ghosh et al.
Photochemistry and photobiology, 95(1), 293-305 (2018-08-17)
There is a major need for light-activated materials for the release of sensitizers and drugs. Considering the success of chiral columns for the separation of enantiomer drugs, we synthesized an S,S-chiral linker system covalently attached to silica with a sensitizer
Epoxidation of olefins catalyzed by novel Mn(III) and Mo(IV) Salen complexes immobilized on mesoporous silica gel: Part I. Synthesis and characterization of homogeneous and immobilized Mn(III) and Mo(IV) Salen complexes
Luts T, et al.
J. Mol. Catal. A: Chem., 261(1), 16-23 (2007)
Mari Miura Sugii et al.
Materials science & engineering. C, Materials for biological applications, 73, 340-346 (2017-02-12)
The antibiofilm effect of iodide quaternary ammonium methacryloxy silicate (IQAMS) in Transbond XT Light Cure Adhesive resin used for braces cementation was evaluated. Fourier Transform Infrared (FTIR) spectroscopy confirmed IQAMS formation and Scanning Electron Microscopy coupled to Energy-Dispersive X-ray Spectroscopy
Sandra Jimenez-Falcao et al.
Journal of materials chemistry. B, 7(30), 4669-4676 (2019-08-01)
We report herein the assembly of an integrated nanodevice with bi-enzymatic cascade control for on-command cargo release. This nanocarrier is based on Au-mesoporous silica Janus nanoparticles capped at the mesoporous face with benzimidazole/β-cyclodextrin-glucose oxidase pH-sensitive gate-like ensembles and functionalized with
Organically modified silica nanoparticles with intraparticle heavy-atom effect on the encapsulated photosensitizer for enhanced efficacy of photodynamic therapy.
Kim S, et al.
The Journal of Physical Chemistry C, 113(29), 12641-12644 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service