Skip to Content
Merck
All Photos(1)

Key Documents

T61204

Sigma-Aldrich

Triethyl phosphite

98%

Synonym(s):

P(EtO)3, P(OEt)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3P
CAS Number:
Molecular Weight:
166.16
Beilstein:
956578
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: Reductions

impurities

≤1% triethyl phosphate and diethyl phosphite

refractive index

n20/D 1.413 (lit.)

bp

156 °C (lit.)

density

0.969 g/mL at 25 °C (lit.)

SMILES string

CCOP(OCC)OCC

InChI

1S/C6H15O3P/c1-4-7-10(8-5-2)9-6-3/h4-6H2,1-3H3

InChI key

BDZBKCUKTQZUTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as a reducing agent; can react with electrophiles to form phosphonates or phosphates; forms a stable complex with copper(I) iodide.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stowell, J. K.; Widlanski, T. S. et al.
Tetrahedron Letters, 36, 1825-1825 (1995)
Gabriele Albertin et al.
Dalton transactions (Cambridge, England : 2003), 44(35), 15470-15480 (2015-08-04)
Diazoalkane complexes [Ru(Tp)(N2CAr1Ar2)(PPh3)L]BPh4 ( and ) [Tp = tris(pyrazolyl)borate; L = P(OMe)3, P(OEt)3; Ar1 = Ar2 = Ph; Ar1 = Ph, Ar2 = p-tolyl; Ar1Ar2 = C12H8] were prepared by allowing chloro-compounds RuCl(Tp)(PPh3)L to react with diazoalkane in the presence
A Useful Synthesis of 1-Aminocyclopropanephosphonic Acid from Cyclopropanone Acetal.
Antoine Fadel
The Journal of organic chemistry, 64(13), 4953-4955 (2001-10-25)
Kyriacos Agapiou et al.
Journal of the American Chemical Society, 126(14), 4528-4529 (2004-04-09)
Exposure of enone substrates 1a-18a, which possess appendant ketone, ester, and nitrile moieties, to Et2Zn in the presence of catalytic Cu(OTf)2/P(OEt)3 provides the cyclized products in good to excellent yields and diastereoselectivities. These results represent the first use of ketones
Misner, J. W.; Fisher, J. W. et al.
Tetrahedron Letters, 44, 5991-5991 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service