Skip to Content
Merck
All Photos(1)

Key Documents

440183

Sigma-Aldrich

(3-Chloropropyl)trimethoxysilane

≥97%

Synonym(s):

δ-Chloropropyltrimethoxysilane, (γ-Chloropropyl)trimethoxysilane, (3-Chloropropyl)trimethoxysilane, 3-(Trimethoxysilyl)propyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
198.72
Beilstein:
969627
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥97%

form

liquid

refractive index

n20/D 1.419 (lit.)

bp

195 °C/750 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCCl)(OC)OC

InChI

1S/C6H15ClO3Si/c1-8-11(9-2,10-3)6-4-5-7/h4-6H2,1-3H3

InChI key

OXYZDRAJMHGSMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(3-Chloropropyl)trimethoxysilane (CPTMS) is an organo-silane which forms a self-assembled monolayer (SAMs) that facilitates the surface modification of different bio-and nano- materials.

Application

(3-Chloropropyl)trimethoxysilane may be used in the synthesis of octakis(3-chloropropyl)octasilsesquioxane.{35}
CPTMS can be used to silanize a variety of nanoparticles like carbon nanotubes (CNT) and silica nanoparticles (SiO2) for different applications like development for molecularly imprinted polymers for protein detection and fabrication of mesoporous silica(organic-inorganic hybrid). It may be used to functionalize silica-iron oxide nanoparticles which can be used as catalysts for the synthesis of pyrano(3,2−c:5,6−c′)dichromene-6,8-dione derivatives.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.1 °F - closed cup

Flash Point(C)

71.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marie-Alexandra Neouze et al.
Monatshefte fur chemie, 143(4), 519-525 (2012-01-01)
A newly arising challenge in the field of nanoparticle research concerns the control and the understanding of the interparticle interactions and interparticle properties. This should allow the development of materials based on nanoparticle assemblies which represents a great opportunity to
One-pot synthesis and modification of silica nanoparticles with 3-chloropropyl-trimethoxysilane assisted by microwave irradiation.
De Conto JF, et al.
Chemical Engineering Communications , 205(4), 533-537 (2018)
Durali Mendil et al.
Food chemistry, 221, 1394-1399 (2016-12-17)
A novel and simple solid phase extraction method was improved and recommended for selenium. Silica gel was modified with 2,6-diamino-4-phenil-1,3,5-triazine and characterized by FTIR, SEM and elemental analysis and used adsorbent for column solid phase extraction of selenium ions. The
Relation between the wetting effect and the adsorbed amount of water-soluble polymers or proteins at various interfaces.
Pandey LM and Pattanayek SK
Journal of Chemical and Engineering Data, 58(12), 3440-3446 (2013)
Investigation on the dielectric properties of titanium-loaded ionic liquid-based nano-organosilica as a novel material.
Vaziri B, et al.
Advanced Powder Technology, 29(3), 813-817 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service