Skip to Content
Merck
All Photos(1)

Key Documents

91877

Sigma-Aldrich

Trimethoxy(3,3,3-trifluoropropyl)silane

≥97.0% (GC)

Synonym(s):

3,3,3-Trifluoropropyl-trimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13F3O3Si
CAS Number:
Molecular Weight:
218.25
Beilstein:
1762962
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.355

bp

144 °C (lit.)

density

1.142 g/mL at 20 °C (lit.)

functional group

fluoro

SMILES string

CO[Si](CCC(F)(F)F)(OC)OC

InChI

1S/C6H13F3O3Si/c1-10-13(11-2,12-3)5-4-6(7,8)9/h4-5H2,1-3H3

InChI key

JLGNHOJUQFHYEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethoxy(3,3,3-trifluoropropyl)silane is an organosilicon compound used as a reagent to synthesize various organosilicon compounds.

Application

Trimethoxy(3,3,3-trifluoropropyl)silane is commonly used in the synthesis of a variety of silsesquioxane based polymers. It is a precursor in the synthesis of macroporous gels for separation of oil and water. It can also be used in the synthesis of hydrophobic silica aerogel for the removal of uranium from aqueous solutions.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precision synthesis of a fluorinated polyhedral oligomeric silsesquioxane-terminated polymer and surface characterization of its blend film with poly (methyl methacrylate).
Koh K, et al.
Macromolecules, 38(4), 1264-1270 (2005)
Granulated activated carbon modified with hydrophobic silica aerogel-potential composite materials for the removal of uranium from aqueous solutions.
Coleman S J, et al.
Environmental Science & Technology, 37(10), 2286-2290 (2003)
Facile synthesis of marshmallow?like macroporous gels usable under harsh conditions for the separation of oil and water.
Hayase G, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 125(7), 2040-2043 (2013)
Hepta (3, 3, 3-trifluoropropyl) polyhedral oligomeric silsesquioxane-capped poly (N-isopropylacrylamide) telechelics: synthesis and behavior of physical hydrogels.
Wang L, et al.
ACS Applied Materials & Interfaces, 3(3), 898-909 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service