Skip to Content
Merck
All Photos(1)

Key Documents

470775

Sigma-Aldrich

Vanadium(V) oxytriethoxide

95%

Synonym(s):

Ethyl orthovanadate, Oxovanadium triethoxide, Tri(ethoxy)vanadium oxide, Triethoxyvanadyl, Triethyl vanadate, Vanadium oxytriethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OV(OC2H5)3
CAS Number:
Molecular Weight:
202.12
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

reaction suitability

core: vanadium
reagent type: catalyst

bp

125-128 °C (lit.)

density

1.139 g/mL at 25 °C (lit.)

SMILES string

CCO[V](=O)(OCC)OCC

InChI

1S/3C2H5O.O.V/c3*1-2-3;;/h3*2H2,1H3;;/q3*-1;;+3

InChI key

IOHMVQGDVUCILM-UHFFFAOYSA-N

Application

Building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands the complexes have potential biological relevance as well as uses in catalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feilong Jiang et al.
Inorganic chemistry, 37(21), 5439-5451 (2001-10-24)
The studies of an underexplored synthetic reagent, VO(2)(acac) (Hacac = acetylacetone) and semirational strategies for the formation of a complete series of simple vanadium(V) alkoxide clusters in alcohol-containing solvents. The neutral mono-, di-, tri-, and tetranuclear oxovanadium(V) complexes [V(2)O(2)(&mgr;-OCH(3))(2)(acac)(2)(OCH(3))(2)] (1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service