Skip to Content
Merck
All Photos(4)

Key Documents

289957

Sigma-Aldrich

Sodium dodecylbenzenesulfonate

technical grade

Synonym(s):

SDBS, Dodecylbenzenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11C6H4SO3Na
CAS Number:
Molecular Weight:
348.48
Beilstein:
4171051
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

powder

mp

>300 °C (> 572 °F)

solubility

water: soluble 0,8  g/L °C

density

1 g/cm3 at at-20 °C (68 °F)

functional group

sulfonic acid

SMILES string

[Na+].CCCCCCCCCCCCc1ccc(cc1)S([O-])(=O)=O

InChI

1S/C18H30O3S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-17-13-15-18(16-14-17)22(19,20)21;/h13-16H,2-12H2,1H3,(H,19,20,21);/q;+1/p-1

InChI key

JHJUUEHSAZXEEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium dodecylbenzenesulfonate (SDBS) acts as a surfactant in the ionic self-assembly reaction of cyclophane BIMCP-1.
Sodium dodecylbenzenesulfonate is an ionic surfactant.

Application

Sodium dodecylbenzenesulfonate has been used to stabilize dispersions of graphene nanoflakes (GNFs) during the preparation of the liquid phase of GNFs. It can also suspend single-walled carbon nanotubes as individuals in aqueous media and also give well-resolved spectral features.
Sodium dodecylbenzenesulfonate was used to fabricate amperometic biosensors for glucose.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinglong Pan et al.
ACS applied materials & interfaces, 13(24), 28864-28869 (2021-06-09)
Polymer composites have attracted increasing interest as thermal management materials for use in devices owing to their ease of processing and potential lower costs. However, most polymer composites have only modest thermal conductivities, even at high concentrations of additives, resulting
Low operation voltage macromolecular composite memory assisted by graphene nanoflakes.
Lai YC, et al.
Journal of Material Chemistry C, 1(3), 552-559 (2013)
Individually suspended single-walled carbon nanotubes in various surfactants.
Moore VC, et al.
Nano Letters, 3(10), 1379-1382 (2003)
Jie Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 29(33), 10586-10595 (2013-07-19)
Electrodes modified with single-walled carbon nanotubes (SWNTs) offer a number of attractive properties for developing novel electrochemical sensors. A common method to immobilize SWNTs onto the electrode surface is by placing a droplet of a SWNT suspension onto the electrode
Sandeep Kumar et al.
Organic letters, 15(13), 3400-3403 (2013-06-22)
Cyclophane BIMCP-1 undergoes ionic self-assembly with surfactant sodium dodecylbenzenesulfonate (SDBS) at a concentration far below its CMC value to form aggregates with spherical morphology. The rotational restriction of rings in these aggregates facilitates 32-fold enhancement in emission intensity (AIEE) to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service