Skip to Content
Merck
All Photos(5)

Key Documents

208272

Sigma-Aldrich

Vanadium(III) chloride

97%

Synonym(s):

Vanadium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
VCl3
CAS Number:
Molecular Weight:
157.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: vanadium

density

3 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[V](Cl)Cl

InChI

1S/3ClH.V/h3*1H;/q;;;+3/p-3

InChI key

HQYCOEXWFMFWLR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Vanadium(III) chloride is a crystalline solid with variable oxidation states. It is widely used as a catalyst in organic synthesis, polymerization processes, and redox batteries. It acts as a reducing agent.

Application

Vanadium chloride may be used as a reduction agent to determine nitrates and nitrites in water samples.It may be used in redox flow batteries. Vanadium trichloride catalyses Pinacol coupling reactions of benzaldehyde in water. it also acts as a mild and efficient catalyst for the chemoselective deprotection of acetonides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xiaoliang Xu et al.
The Journal of organic chemistry, 70(21), 8594-8596 (2005-10-08)
A catalytic pinacol coupling using water as a solvent was performed by a catalytic amount of vanadium(III) chloride and metallic Al as a co-reductant. A combination forms a binary catalytic system, being in sharp contrast to the reaction in organic
Thanh-Binh Nguyen et al.
Environmental management, 63(4), 455-465 (2018-02-06)
The objective of this study was to examine the feasibility of sewage sludge composting using a simple aeration method. Two consecutive composting trials (run A and run B) using Japanese sludge and woodchips (1:1, v/v) were conducted in cubic boxes
Heyam Mohamed Ali Sidahmed et al.
BMC complementary and alternative medicine, 13, 183-183 (2013-07-23)
Mitrella kentii (M. kentii) (Bl.) Miq, is a tree-climbing liana that belongs to the family Annonaceae. The plant is rich with isoquinoline alkaloids, terpenylated dihydrochalcones and benzoic acids and has been reported to possess anti-inflammatory activity. The purpose of this
Lingjun Kong et al.
Small (Weinheim an der Bergstrasse, Germany), 14(21), e1800639-e1800639 (2018-04-20)
Hierarchical porous structures are highly desired for various applications. However, it is still challenging to obtain such materials with tunable architectures. Here, this paper reports hierarchical nanomaterials with oriented 2D pores by taking advantages of thermally instable bonds in vanadium-based
Novel vanadium chloride/polyhalide redox flow battery.
Skyllas-Kazacos M
Journal of Power Sources, 124(1), 299-302 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service