Skip to Content
Merck
All Photos(2)

Key Documents

339113

Sigma-Aldrich

Tantalum(V) ethoxide

99.98% trace metals basis

Synonym(s):

Pentaethoxytantalum, Pentaethyl tantalate, Tantalum ethylate, Tantalum pentaethoxide, Tantalum(V) ethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ta(OC2H5)5
CAS Number:
Molecular Weight:
406.25
Beilstein:
3678999
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.98% trace metals basis

form

liquid

reaction suitability

core: tantalum
reagent type: catalyst

impurities

<2 wt. % toluene

refractive index

n20/D 1.487 (lit.)

bp

155 °C/0.01 mmHg (lit.)

mp

21 °C (lit.)

density

1.566 g/mL at 25 °C (lit.)

SMILES string

CCO[Ta](OCC)(OCC)(OCC)OCC

InChI

1S/5C2H5O.Ta/c5*1-2-3;/h5*2H2,1H3;/q5*-1;+5

InChI key

HSXKFDGTKKAEHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tantalum(V) ethoxide Ta(OC2H5)5 is a tantalum alkoxide that can be used as a precursor in the preparation of tantalum oxide by chemical vapor deposition (CVD). It has excellent volatility and thermal stability. It can be synthesized by anodic oxidation of tantalum plate in anhydrous ethanol.

Application

Ta(OC2H5)5 can be used in the formation of ultrathin tantalum oxide based capacitors. It can also be used as a liquid precursor in the preparation of tantalum carbide nanopowder for hard coating on metal surfaces.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Electrosynthesis of Tantalum Ethoxide
Yang H, et al.
Electrochemistry, 82(9), 743-748 (2014)
Leakage currents in amorphous Ta 2 O 5 thin films
Chiu F, et al.
Journal of Applied Physics, 81(10), 6911-6915 (1997)
Controlling the synthesis of TaC nanopowders by injecting liquid precursor into RF induction plasma
Ishigaki T, et al.
Science and Technology of Advanced Materials, 6(2), 111-111 (2005)
Ultrathin tantalum oxide capacitor dielectric layers fabricated using rapid thermal nitridation prior to low pressure chemical vapor deposition
Kamiyama S, et al.
Journal of the Electrochemical Society, 140(6), 1617-1625 (1993)
Kuo-Shyan Lin et al.
Cancer research, 75(2), 387-393 (2014-12-10)
The bradykinin receptor B1R is overexpressed in many human cancers where it might be used as a general target for cancer imaging. In this study, we evaluated the feasibility of using radiolabeled kallidin derivatives to visualize B1R expression in a

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service